TSX Series High-Performance -30°C Auto Defrost Freezers
TSX Series High-Performance -30°C Auto Defrost Freezers
TSX Series High-Performance -30°C Auto Defrost Freezers
TSX Series High-Performance -30°C Auto Defrost Freezers
TSX Series High-Performance -30°C Auto Defrost Freezers
Thermo Scientific™

TSX Series High-Performance -30°C Auto Defrost Freezers

Thermo Scientific™ TSX Series high-performance -30°C auto defrost freezers offer a sustainable choice for clinical and laboratory storage.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacity (English)Plug TypeElectrical Requirements
TSX2330FA23 cu. ft.NEMA 5-15115V, 60Hz
TSX5030FA51.1 cu. ft.NEMA 5-15115V, 60Hz
TSX5030FD51.1 cu. ft.NEMA 6-15208-230V, 60Hz
TSX5030FV51.1 cu. ft.CEE 7/7208-230V, 50Hz
TSX1230FV11.5 cu. ft.CEE 7/7208-230V, 50Hz
TSX1230FY11.5 cu. ft.NEMA 5-15115 V 60 Hz
TSX1230FZ11.5 cu. ft.NEMA 6-15208 - 230 V 60 Hz
TSX3030FV29.2 cu. ft.CEE 7/7208-230V, 50Hz
TSX3030FY29.2 cu. ft.NEMA 5-15115 V 60 Hz
TSX3030FZ29.2 cu. ft.NEMA 6-15208 - 230 V 60 Hz
TSX5030FY51.1 cu. ft.NEMA 5-20115 V 60 Hz
TSX2330FD23 cu. ft.NEMA 6-15208-230V, 50/60Hz
TSX5030FZ51.1 cu. ft.NEMA 6-15208 - 230 V 60 Hz
TSX2330FV23 cu. ft.CEE 7/7208-230V, 50/60Hz
TSX2330FY23.3 cu. ft.NEMA 5-15115 V 60 Hz
TSX2330FZ23.3 cu. ft.NEMA 6-15208 - 230 V 60 Hz
Catalog number TSX2330FA
Price (INR)
-
Capacity (English):
23 cu. ft.
Plug Type:
NEMA 5-15
Electrical Requirements:
115V, 60Hz

Thermo Scientific™ TSX Series high-performance -30°C auto defrost freezers are designed with features that support sample protection and sustainability objectives for the storage reagents, pharmaceuticals, vaccines, biological and other commonly used laboratory and clinical materials such as primers, siRNA molecules and fetal bovine serum (FBS) requiring -30°C. The Series' V-drive technology is designed to provide temperature uniformity that continually adapts to user patterns, offering significant energy savings without compromising protection.

Certified to NSF/ANSI 456 Vaccine Storage* standard, the TSX series can help you meet regulations and guidelines from the CDC, World Health Organization, and other Health Ministries on vaccine storage and handling.

The TSX Series delivers the performance and quality you need, in addition to the low sound, low energy, and exceptional user experience you deserve.

*For NSF/ANSI 456 compliance, units must be ordered with a certificate of calibration (6916-2)

The drive for sample integrity and security

  • Designed to safely store media, reagents, pharmaceuticals, vaccines, chemotherapy and other laboratory and medical-grade storage requiring a tight temperature performance
  • Unique V-drive technology is designed to detect usage patterns such as door openings when a higher compressor speed is needed and periods of stability where the compressor runs at a lower speed, saving energy without compromising protection
  • Non-invasive fully adaptive defrost with time and temperature guided cycle
  • Easy to use microprocessor control with interior light, setpoint security and alarms
  • Standard door locks provide comfort and assurance

Enabling compliance

  • Tested and Certified to NSF/ANSI 456 Vaccine Storage standard and supports the US CDC, WHO, and other global vaccine storage guidelines and recommendations*
  • Tested to GMP Clean Room Class A / ISO 5 & 6 (ISO EN 14644-1) with easy to clean surfaces support USP <797> and <800> hazardous drug compounding and other clean room environments
  • Tested and certified to conform with the design and safety standards of UL 471, 61010, and CE Directive 2011/65/EU and 2015/863

Designed for an ergonomic and comfortable experience

  • Ergonomically designed door handle with a soft-touch grip
  • The controller is angled to help prevent neck strain
  • Easy to clean and adjust epoxy coated shelves
  • Whisper quiet with a sound level of 52 dBA, enabling end users to work alongside the refrigerator without distraction
  • Self-closing door with 90° stop to assist with inventory loads

Verified sustainability

  • ACT Environmental Impact Factor Label Certified, providing third-party verification of the sustainable impacts of the product, its operations, and its end of life.
  • Natural, R290 refrigerants and water-blown foam make the TSX Series compliant with EU F-GAS regulations, U.S. Environmental Protection Agency's Significant New Alternatives Policy (SNAP), and other Global Environmental Regulations.
  • Manufactured in an award-winning, zero-waste to landfill facility

Ready for connectivity and monitoring

  • Thermo Scientific™ DeviceLink™ Connect ready, enabling you to have a connected experience with your unit so you always—and anywhere— know the safety of your samples and the performance of your refrigerator or freezer.
  • Compatible with wireless monitoring systems and remote alarm contacts including our Thermo Scientific™ Smart-Vue™ Pro wireless monitoring systems

Make it your own

  • Alternate Door swing configuration and door types
  • Extra and Alternate wire baskets, shelves, and drawers
  • Free-standing or factory-installed chart recorders
  • Stainless Steel interior finish
  • E-Lock Adapter kits
  • 4-20 milliamp transmitters and other remote alarm modules
  • Specialty, regional, and country specific plugs

Services and support

  • Industry Leading 120-month compressor warranty (varies by country, please contact us for details)
  • Optional Certificate of Calibration to an NIST (National Institute of Standards and Technology, formerly National Bureau of Standards) traceable instrument includes a model number and serial number certificate
  • Optional Validation Testing Programs include in house and field-based temperature mapping and Installation Qualification (IQ) and Operation Qualification (OQ) programs
  • Optional Factory Acceptance Test results including QC Test Log Sheets and QC Temperature Graphs

*For NSF/ANSI 456 compliance, units must be ordered with a certificate of calibration (6916-2)

** Data on file. Based on comparisons between TSX Series and previous generation Thermo Scientific models.

Data on file. Based on comparisons between TSX Series and previous generation Thermo Scientific models.
Specifications
Amperage15 A
Chart RecorderOptional
DefrostAutomatic
Depth (English) Exterior37.8 in.
Depth (English) Interior28.5 in.
Depth (Metric) Exterior96 cm
Depth (Metric) Interior72.3 cm
Dimensions (D x W x H) Interior28.5 x 24 x 58 in. (72.3 x 61 x 147.3 cm)
Dimensions (L x W x H) Exterior37.8 x 28 x 78.6 in. (96 x 71.1 x 199.6 cm)
DisplayMicroprocessor
Door StyleSolid
Electrical Requirements115V, 60Hz
Frequency60 Hz
Height (English) Exterior78.6 in.
Height (English) Interior58 in.
Height (Metric) Exterior199.6 cm
Height (Metric) Interior147.3 cm
Plug TypeNEMA 5-15
Product LineTSX Series
Rack Capacity8
RefrigerantR290
Shipping Weight (English)423 lb.
Shipping Weight (Metric)198 kg
Smart View CompatibilitySmart-Vue Compatible
Temperature (Setpoint)-35° to -15°C (-30°C)
TypeHigh-Performance Freezer
WarrantyWarranty varies by country, please contact us for details
Weight (English)358 lb.
Weight (Metric)162 kg
Width (English) Exterior28 in.
Width (English) Interior24 in.
Width (Metric) Exterior71.1 cm
Width (Metric) Interior61 cm
Capacity (English)23 cu. ft.
Capacity (Metric)650 L
Certifications/ComplianceUL, cUL, NSF 456 (When Purchased with 6916-2 Calibration Certificate)
CompressorV-drive
Monitoring OptionsWireless
No. of Doors1
Shelves4
StyleUpright
Temperature Range-15°C to -35°C
Voltage115 V
Unit SizeEach

Frequently asked questions (FAQs)

Can the TSX Series High-Performance -30 degrees C Auto Defrost Freezers be locked?

Yes, The TSX Series freezers come with standard door locks to offer comfort and assurance, helping ensure the security of the stored materials.

Additionally, the units are equipped with a key-operated triple position switch for main power and alarm, which locks in temperature and alarm setpoints to minimize setpoint error and prevent tampering.

For enhanced access control, E-lock adapter kits are available. These E-locks provide access control with audit-trail capabilities and temperature monitoring. They don't require electricity and are available in both factory- and customer-installed options for various lock systems such as CompX™, Pyxis™, Omnicell™, and McKesson™locks. For additional details, refer to the TSX Series High-Performance Refrigerators and Freezers Brochure.