MAS™ T-Marker™ Controls
Actual product may vary
Thermo Scientific™
MAS™ T-Marker™ Controls
Monitor the performance of various cancer antigen tests on automated instruments with liquid assayed, multi-analyte Thermo Scientific™ MAS™ T-Marker Controls.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberLevel
TUM-3033
TUM-2022
TUM-1011
Catalog number TUM-303
Price (INR)
-
Level:
3
Monitor the performance of various cancer antigen tests on automated instruments with liquid assayed, multi-analyte Thermo Scientific™ MAS™ T-Marker Controls. In compliance with NACB and EGTM* guidelines, MAS T-Marker Quality Controls allow for independent evaluation of test performance in your lab. With 26 analytes per vial, a 59-month shelf life, and ready-to-use design, you are free to focus on other tasks for years following lot rollover.
  • 59 months industry-leading shelf life
  • Liquid, ready to use; no additional preparation required
  • Human serum matrix
  • Includes CA 15-3, CA 19-9, CA 72-4, CA-125, CA 27-29
  • Assayed values available for most major analyzers.
  • 26 analytes per vial: ACTH*, Aldosterone, AlphaFetoprotein (AFP), Beta Human Chorionic Gonadotroopin, (beta-hCG), Beta-2 Microglobulin, CA 125, CA 15-3,CA 19-9, CA 27-29, CA 72-4, Calcitonin, Carcinoembryonic Antigen (CEA), Cortisol, CyFR A 21-1*, Estradiol, Ferritin, Gastrin, Human Chorionic Gonadotroopin (hCG),  Insulin,  NSE*,  Prolactin, Prostate Specific Antigen, Total (TPSA), Prostate Specific Antigen, Free (FPSA), Prostatic Acid Phosphatase (PAP), Thyroglobulin
*Not cleared by FDA. These constituent levels have not been adjusted but are at levels found in the source material used in preparation of the controls. No claim is made for expected values of these constituents.
For In Vitro Diagnostic Use.  
For In Vitro Diagnostic Use.  
Specifications
DescriptionTumor Marker Control, Assayed
Level3
Packaging6 Pk.
Quantity3 mL
TypeControl
Detectable AnalytesAdrenocorticotropic Hormone (ACTH), alpha-Fetoprotein (AFP), Aldosterone, CA 15-31, CA 19-91, CA 72-41, CA-125, CA-29, Calcitonin, Carcinoembryonic Antigen (CEA), Cortisol, Estradiol, Ferritin, Gastrin, hCG, β-hCG, Insulin, beta-2-microglobulin (B2M), Prostatic Acid Phosphatase (PAP), Prolactin, PSA, PSA (Free), Thryoglobulin
Sample TypeSerum
Unit SizeEach
Contents & Storage
59-month shelf life @ -25 to -15 °C from date of manufacture
90 days unopened @ 2 to 8 °C
30 days open vial @ 2 to 8 °C