Differentiate members of the family Enterobacteriaceae based on whether bacteria can utilize citrate as their sole carbon source, indicated by a color change in the medium.
Have Questions?
Catalog Number
Fill Weight
TV5015Z
7 to 8 g
Catalog number TV5015Z
Price (INR)
-
Fill Weight:
7 to 8 g
Simmons Citrate Agar (Slant) is used to differentiate members of the family Enterobacteriaceae based on whether bacteria can utilize citrate as their sole carbon source, indicated by a color change in the medium.