Lysine Decarboxylase Broth (Taylor)
Lysine Decarboxylase Broth (Taylor)
Thermo Scientific™

Lysine Decarboxylase Broth (Taylor)

Detect lysine decarboxylase enzyme activity in by salmonellae and other Enterobacteriaceae with this medium.
Have Questions?
Catalog NumberFill Weight
TV5028N5.5 to 6.5 g
Catalog number TV5028N
Price (INR)
-
Request A Quote
Fill Weight:
5.5 to 6.5 g
Detect lysine decarboxylase enzyme activity in by salmonellae and other Enterobacteriaceae with Lysine Decarboxylase Broth (Taylor), which helps differentiate enteric bacteria based on their ability to decarboxylate lysine and produce cadaverine.
Specifications
ColorPearl Violet, Transparent
Fill Weight5.5 to 6.5 g
FormatPoured Tube
Packaging Type50 Tubes/Box
Quantity50/Pk.
Shelf Life32 weeks
Manufacturing OriginGermany
Product TypeMicrobiology Media
Unit SizeEach
Contents & Storage
2°C to 12°C