Muller-Kauffmann Tetrathionate-Novobiocin (MkTTn) Enrichment Broth
Thermo Scientific™
Muller-Kauffmann Tetrathionate-Novobiocin (MkTTn) Enrichment Broth
Selectively enrich Salmonella species from food with the convienience of Muller-Kauffmann Tetrathionate-Novobiocin Enrichment Broth prepared media tubes.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberFill WeightManufacturing Origin
TV5065E10 ± 1mLGermany
BO1224K50 to 51.5 gUnited Kingdom
Catalog number TV5065E
Price (INR)
-
Fill Weight:
10 ± 1mL
Manufacturing Origin:
Germany
Selectively isolate Salmonella species with the convenience of Thermo Scientific™ Muller-Kauffmann Tetrathionate-Novobiocin (MkTTn) Enrichment Broth prepared media tubes. MkTTn is a selective enrichment medium for the isolation of Salmonella, whose formulation conforms to ISO 6579:20021 for the microbiological testing food and animal feeding stuffs.
Muller-Kauffmann Tetrathionate-Novobiocin Broth was developed by Muller2 and later modified by Kauffmann3,4 with the addition of ox bile and brilliant green to improve selectivity. The addition of novobiocin at 40 mg per liter was later described by Jeffries5 to improve inhibition of Proteus species.
After the sample has been incubated in Buffered Peptone Water (ISO), transfer 1 mL of the broth to 10 mL of MkTTn Broth and 0.1 mL into 10 mL of RVS Broth (TV5036E). Incubate the MkTTn Broth at 37°C ± 1°C for 24 hours ± 3 hours and the RVS Broth at 41.5°C ± 1°C, for 24 hours ± 3 hours. Sub-culture the incubated MkTTn and RVS broths onto XLD Agar (PO5057A/PO0164A) and a second agar medium of choice, and incubate for a further 24 hours ± 3 hours at 37°C ± 1°C. Presumptive Salmonella should be confirmed using appropriate biochemical and serological techniques. For the complete method, please refer to ISO 6579:20021.
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand, combining powerful manual, semi-automated and fully automated test products and a comprehensive line of media and diagnostic products to offer a complete, end-to-end solution to quickly deliver the products you need and the quality results your laboratory depends on.
For laboratory use only.
General References:
  1. Anon BS EN ISO 6579:2002. Microbiology of food and animal feeding stuffs. Horizontal method for the detection of Salmonella spp.
  2. Muller L. (1923) C. R. Soc. Biol (Paris) 89. 434-447.
  3. Kauffmann F. (1930) Z. f. Hyg. 113. 148-152.
  4. Kauffmann F. (1935) Z. f. Hyg. 117. 26-32.
  5. Jeffries L. (1959) J. Clin. Path. 12. 568-571.
Specifications
ColorLight green, opaque
DescriptionMuller Kauffmann Tetrathionate Novobiocin Enrichment Broth
Fill Volume10 mL
Fill Weight10 ± 1mL
FormatTube
Packaging Type50 tubes in a carton box
Quantity50/Pk.
Shelf Life12 Weeks
Manufacturing OriginGermany
Product TypeMicrobiology Media
Unit SizeEach
Contents & Storage
2°C to 12°C