Tryptone Soya Broth with Tween™ 80
Thermo Scientific™

Tryptone Soya Broth with Tween™ 80

Recover and enumerate microorganisms from samples, especially where disinfectant neutralization or dispersion of hydrophobic organisms is needed during microbiological testing.
Have Questions?
Catalog NumberFill Weight
TV5086D8.5 to 9.5 g
Catalog number TV5086D
Price (INR)
-
Fill Weight:
8.5 to 9.5 g
Tryptone Soya Broth with Tween™ 80 is used to recover and enumerate microorganisms from samples, especially where disinfectant neutralization or dispersion of hydrophobic organisms is needed during microbiological testing.
Specifications
ColorMaize Yellow, Transparent
Fill Weight8.5 to 9.5 g
FormatPoured Tube
Packaging Type50 Tubes/Box
Quantity50/Pk.
Shelf Life52 Weeks
Manufacturing OriginGermany
Product TypeMicrobiology Media
Unit SizeEach
Contents & Storage
2°C to 25°C