Todd Hewitt Broth with Colistin and Nalidixic acid (LIM Broth)
Thermo Scientific™

Todd Hewitt Broth with Colistin and Nalidixic acid (LIM Broth)

Cultivate and enrich fastidious organisms, particularly Streptococci and Group B Streptococcus (GBS), from clinical or food samples.
Have Questions?
Catalog NumberFill Weight
TV5093E4.5 to 5.5 g
Catalog number TV5093E
Price (INR)
-
Fill Weight:
4.5 to 5.5 g
Todd-Hewitt Broth is ready to use medium originally described by Todd and Hewitt for the production of antigenic streptococcal haemolysin.
Specifications
ColorSand Yellow, Transparent
Fill Weight4.5 to 5.5 g
FormatPoured Tube
Packaging Type50 Tubes/Box
Quantity50/Pk.
Shelf Life10 Weeks
Manufacturing OriginGermany
Product TypeMicrobiology Media
Unit SizeEach
Contents & Storage
2°C to 12°C