Selectively isolate and enumerate Clostridium perfringens in food and water samples by detecting sulfite reduction, which produces black colonies under anaerobic conditions.
Have Questions?
Catalog Number
Fill Weight
TV5204G
19.5 to 20.5 g
Catalog number TV5204G
Price (INR)
-
Fill Weight:
19.5 to 20.5 g
TSC Agar Base is used to selectively isolate and enumerate Clostridium perfringens in food and water samples by detecting sulfite reduction, which produces black colonies under anaerobic conditions.