Search
Search

| Catalog Number | Quantity | Format |
|---|---|---|
| TWT9005A | (5x68) 340 plates | 90mm settle plate |
| TWT5505A | (5x100) 500 plates | 55mm contact plate |
| TWT9005AF | (5x10) 50 plates | 90mm settle plate |
Experience next-level cleanroom and isolator confidence with LabServ™ TSA with Lecithin and Polysorbate 80 TWI Plates. The irradiated plates, triple wrapped in transparent packaging, are designed to meet the stringent requirements of environmental monitoring in the pharmaceutical industry.
Reduce the risk of contamination for confidence in your results with sealed, sterile and secure high-quality Oxoid™ culture media plates. Quantitative quality control testing ensures optimum performance in accordance with British1, European2, Japanese3 and US4 Pharmacopoeia.
Streamline environmental monitoring of cleanroom and isolator environments with Tryptic Soya Agar with Lecithin and Polysorbate 80 TWI Plates.
Not all products are available for sale in all territories. Please inquire.
General References: