Stainless Steel Unions, Tees and Crosses
Thermo Scientific™
Stainless Steel Unions, Tees and Crosses
Minimize system dead volumes and ensure reliable high-pressure connections in your HPLC system with Stainless-Steel Unions, Tees and Crosses.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberTypeDescriptionSwept Volume
U-428Tee0.020 in. through hole0.57 μL
U-402Union0.020 in. through hole0.134 μL
U-412Union0.020 in. through hole0.129 μL
Catalog number U-428
Price (INR)
-
Type:
Tee
Description:
0.020 in. through hole
Swept Volume:
0.57 μL
Minimize system dead volumes and ensure reliable high-pressure connections in your HPLC system with Thermo Scientific™ Stainless-Steel Unions, Tees and Crosses. Includes two stainless-steel nuts and ferrules.
  • Absolute zero or low dead volume formats
  • Includes two stainless steel nuts and ferrules
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Specifications
Description0.020 in. through hole
IncludesTee
Swept Volume0.57 μL
For Use With (Equipment)1/16 in. OD Tubing
Length (Metric)22.61 mm
MaterialStainless Steel
TypeTee
Unit SizeEach