MAS™ UA Controls
Actual product may vary
Thermo Scientific™

MAS™ UA Controls

Monitor urine dipstick and microscopic components with bi-level controls with improved room temperature storage capability.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberDescriptionLevelQuantity
UAB-260UA Controls, Assayed260 mL
UAB-115UA Controls, Assayed115 mL
UAB-215UA Controls, Assayed2 (abnormal)15 mL
UAB-MPUA Controls, Assayed, Bi-levelBi-level15 mL
UAB-160UA Controls, Assayed160 mL
Catalog number UAB-260
Price (INR)
-
Description:
UA Controls, Assayed
Level:
2
Quantity:
60 mL

Monitor urine dipstick and microscopic components with Thermo Scientific™ MAS™ UA Controls, bi-level controls with improved room temperature storage capability. Liquid and ready-to-use, this urine control is assayed for most major reagents strips and strip systems. It includes 10 commonly-assayed dipstick tests plus Microalbumin, Creatinine and hCG.

Ordering Information:
Available in convenient 15mL dropper bottles
For In Vitro Diagnostic Use.  
For In Vitro Diagnostic Use.  
Specifications
DescriptionUA Controls, Assayed
Level2
Packaging4 Pk.
Quantity60 mL
Shelf Life24 months
TypeControl
Detectable AnalytesBilirubin, Blood, Creatinine, Crystals, Glucose, hCG, Ketones, Leukocyte Esterase, Microalbumin, Nitrite, Osmolality, pH, Potassium, Protein, Red Blood Cells, Sodium, Specific Gravity, Urobilinogen, White Blood Cells
Sample TypeUrine
Unit SizeEach
Contents & Storage
2°C to 8°C