Thermoluminescent Dosimetry Powders
Thermo Scientific™

Thermoluminescent Dosimetry Powders

Get accurate radiation dose measurement for any application, from health physics to environmental dosimetry applications.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberFor Use With (Application)Emission Spectra
UNW18020Health and Medical Physics Dosimetry3500 to 6000 Å (4000 maximum)
UCW71727Gamma, Beta and Environmental4000 Å
UNW78020Gamma and Beta Dosimetry3500 to 6000 Å (4000 maximum)
UNW28020Environmental Dosimetry4835 Å
UNW68020Neutron Dosimetry3500 to 6000 Å (4000 maximum)
UCW11727Environmental, Personnel, Extremity Dosimetry4000 Å
UCW61727Neutron Dosimetry4000 Å
Catalog number UNW18020
Price (INR)
-
For Use With (Application):
Health and Medical Physics Dosimetry
Emission Spectra:
3500 to 6000 Å (4000 maximum)

Get accurate radiation dose evaluation for applications from health physics to environmental to personnel and extremity, and beyond with highly sensitive thermoluminescent dosimetry powders. Thermo Scientific™ Thermoluminescent Dosimetry Powders are accurate for x-ray radiation, gamma radiation, beta radiation, neutron radiation, and electron radiation according to choice of material.

TLD-600 materials consist of Lithium Fluoride (Li isotope) LiF:Mg, Ti; suitable for gamma and beta dosimetry applications.


Specifications
DescriptionModel No.TLD-100
For Use With (Application)Health and Medical Physics Dosimetry
Measurement Ranges10 pGy to 10 Gy
Emission Spectra3500 to 6000 Å (4000 maximum)
Energy Response1.25
MaterialLithium Fluoride (Li natural) LiF:Mg. Ti
Product TypeThermoluminescent Dosimetry Material
Sensitivity1
Unit SizeEach