Thermoluminescent Dosimetry Powders
Actual product may vary
Thermo Scientific™

Thermoluminescent Dosimetry Powders

Get accurate radiation dose measurement for any application, from health physics to environmental dosimetry applications.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberFor Use With (Application)Emission Spectra
UNW68020Neutron Dosimetry3500 to 6000 Å (4000 maximum)
UCW71727Gamma, Beta and Environmental4000 Å
UNW78020Gamma and Beta Dosimetry3500 to 6000 Å (4000 maximum)
UNW28020Environmental Dosimetry4835 Å
UCW11727Environmental, Personnel, Extremity Dosimetry4000 Å
UCW61727Neutron Dosimetry4000 Å
UNW18020Health and Medical Physics Dosimetry3500 to 6000 Å (4000 maximum)
Catalog number UNW68020
Price (INR)
-
For Use With (Application):
Neutron Dosimetry
Emission Spectra:
3500 to 6000 Å (4000 maximum)

Get accurate radiation dose evaluation for applications from health physics to environmental to personnel and extremity, and beyond with highly sensitive thermoluminescent dosimetry powders. Thermo Scientific™ Thermoluminescent Dosimetry Powders are accurate for x-ray radiation, gamma radiation, beta radiation, neutron radiation, and electron radiation according to choice of material.

TLD-600 materials consist of Lithium Fluoride (Li isotope) LiF:Mg, Ti; suitable for gamma and beta dosimetry applications.


Specifications
For Use With (Application)Neutron Dosimetry
Measurement Ranges10 pGy to 10 Gy
Radiation TypeNeutron
Emission Spectra3500 to 6000 Å (4000 maximum)
Energy Response1.25
MaterialLithium Fluoride (Li isotope) LiF:Mg, Ti
Product TypeThermoluminescent Dosimetry Material
Sensitivity1
Unit SizeEach