MAS™ Sentry Urinalysis Controls
Thermo Scientific™
MAS™ Sentry Urinalysis Controls
Verify the performance of various hCG test kits with Thermo Scientific™ MAS™ Sentry Urinalysis Controls.
Have Questions?
Catalog NumberQuantity
URN500525 mL
Catalog number URN5005
Price (INR)
-
Quantity:
25 mL
Verify the performance of various hCG test kits with Thermo Scientific™ MAS™ Sentry Urinalysis Controls. MAS Sentry Urinalysis Controls can be stored for 12 months at room temperature and 18 months at 2° to 8°C. They are ideal for a wide variety of urinalysis strips.
Features:
  • Ready-to-use liquid controls require no dilution or reconstitution
  • Level 1 is an abnormal control
  • Level 2 is a normal control with hCG and microalbumin
  • May be used to verify the performance of various hCG test kits
Ordering Information:
Packaged in 2 levels, 2 × 25mL each level per kit
For In Vitro Diagnostic Use.  
For In Vitro Diagnostic Use.  
Specifications
DescriptionBi-level Urine Control
Level2
Packaging2 bottles each level
Quantity25 mL
Shelf Life18 months
TypeControl
Detectable AnalytesBilirubin, Blood, Glucose, hCG, Leukocytes, Ketones, Croalbumin, Nitrite, pH, Protein, Specific Gravity, Urobilinogen
Sample TypeUrine
Unit SizeEach
Contents & Storage
2°C to 8°C