Search
Search
| Catalog Number | For Use With (Equipment) | Voltage |
|---|---|---|
| UVS800DDA-115 | SPD111V, SC210A, SPD121P, SPD131DDA, SC250EXP | 115 V |
| UVS400-115 | SPD111V, SC210A | 115 V |
| UVS400A-230 | SPD111V, SC210A | 230 V |
| UVS800DDA-230 | SPD111V, SC210A, SPD121P, SPD131DDA, SC250EXP | 230 V |
Safely handle and recover solvents with the Thermo Scientific™ Savant™ Universal SpeedVac™ Vacuum System, equipped with an easy to use, wide-mouth glass condensation flask. This single integrated unit combines an oil-free vacuum source and refrigerated vapor trap, and can be connected to any of the concentrators to provide a complete SpeedVac system.
Savant UVS400:
Savant US800DDA:
All the standard features as well as:
Requires:
Thermo Scientific™ CryoCool™ heat transfer fluid (model no. SCC1) must be ordered for use with all refrigerated cold traps, and the universal vacuum source. Please order separately.