Sensititre™ Nephelometer
Sensititre™ Nephelometer
Sensititre™ Nephelometer
Sensititre™ Nephelometer
Sensititre™ Nephelometer
Sensititre™ Nephelometer
Thermo Scientific™

Sensititre™ Nephelometer

Cost-effectively standardize inoculum density with the Thermo Scientific™ Sensititre™ Nephelometer.
Have Questions?
Catalog NumberWeight (Metric)
V30110.8 kg
Catalog number V3011
Price (INR)
-
Weight (Metric):
0.8 kg
Optimize performance and achieve accurate results by standardizing bacterial suspension density with the Thermo Scientific™ Sensititre™ Nephelometer. Designed to rapidly prepare a consistent inoculum density, the easy-to-read LED indicator light offers a quick methodology to produce bacterial suspensions equivalent to the required 0.5 McFarland Standard.

The Sensititre Nephelometer reduces the risk of manual errors associated with targeting numeric values, by guiding the user to increase/decrease the concentration of emulsified test colonies, until a green “in range” result is achieved – indicating that the suspension is ready for addition to the chosen test broth.
Not all products are available for sale in all territories. Please inquire.

VersaTREK™ and Sensititre products are now part of the Thermo Scientific brand, combining powerful manual, semi-automated and fully automated test products and a comprehensive line of media and diagnostic products to offer a complete, end-to-end solution to quickly deliver the products you need and the quality results your laboratory depends on.

Specifications
Depth (English)5.3 in.
Depth (Metric)13.5 cm
DescriptionSensititre Nephelometer
Electrical Requirements8-18VAC/DC; 130mA; 1w
Height (English)3.84 in.
Height (Metric)9.7 cm
Weight (English)1.8 lb.
Weight (Metric)0.8 kg
Width (English)7.5 in.
Width (Metric)19 cm
Product TypeNephelometer
Test TypeMinimum Inhibitory Concentration (MIC)
Unit SizeEach