Varioskan™ LUX Multimode Microplate Reader
Varioskan™ LUX Multimode Microplate Reader
Varioskan™ LUX Multimode Microplate Reader
Varioskan™ LUX Multimode Microplate Reader
Varioskan™ LUX Multimode Microplate Reader
Varioskan™ LUX Multimode Microplate Reader
Varioskan™ LUX Multimode Microplate Reader
Thermo Scientific™

Varioskan™ LUX Multimode Microplate Reader

Analyze a variety of samples with five measurement technologies, including absorbance (UV-Vis), fluorescence intensity, Luminescence, AlphaScreen / AlphaLISA and TRF.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberMeasurement TechnologyReading OptionsNo. of DispensersIncludes
VLBLATD1Absorbance, Fluorescence Intensity, Luminescence, Time-resolved Fluorescence, AlphaScreenTop and Bottom1SkanIt PC Software, Research Edition
VL0000D0Absorbance, Fluorescence IntensityTopNoneSkanIt PC Software, Research Edition
VL0L00D0Absorbance, Fluorescence Intensity, LuminescenceTopNoneSkanIt PC Software, Research Edition
VL0L0TD0Absorbance, Fluorescence Intensity, Luminescence, Time-resolved FluorescenceTopNoneSkanIt PC Software, Research Edition
VLBL00GD0Absorbance, Fluorescence Intensity, LuminescenceTop and BottomNoneGas Module, SkanIt PC Software, Research Edition
VLBL00GD1Absorbance, Fluorescence Intensity, LuminescenceTop and Bottom1Gas Module, SkanIt PC Software, Research Edition
VLB000D0Absorbance, Fluorescence IntensityTop and BottomNoneSkanIt PC Software, Research Edition
VLBL00GD2Absorbance, Fluorescence Intensity, LuminescenceTop and Bottom2Gas Module, SkanIt PC Software, Research Edition
VLBL00D0Absorbance, Fluorescence Intensity, LuminescenceTop and BottomNoneSkanIt PC Software, Research Edition
VLBL0TGD0Absorbance, Fluorescence Intensity, Luminescence, Time-resolved FluorescenceTop and BottomNoneGas Module, SkanIt PC Software, Research Edition
VLBL0TGD1Absorbance, Fluorescence Intensity, Luminescence, Time-resolved FluorescenceTop and Bottom1Gas Module, SkanIt PC Software, Research Edition
VLBLATD0Absorbance, Fluorescence Intensity, Luminescence, Time-resolved Fluorescence, AlphaScreenTop and BottomNoneSkanIt PC Software, Research Edition
VLBL0TGD2Absorbance, Fluorescence Intensity, Luminescence, Time-resolved FluorescenceTop and Bottom2Gas Module, SkanIt PC Software, Research Edition
VLBLATGD0Absorbance, Fluorescence Intensity, Luminescence, Time-resolved Fluorescence, AlphaScreenTop and BottomNoneGas Module, SkanIt PC Software, Research Edition
VLBLATGD1Absorbance, Fluorescence Intensity, Luminescence, Time-resolved Fluorescence, AlphaScreenTop and Bottom1Gas Module, SkanIt PC Software, Research Edition
VLBLATGD2Absorbance, Fluorescence Intensity, Luminescence, Time-resolved Fluorescence, AlphaScreenTop and Bottom2Gas Module, SkanIt PC Software, Research Edition
VLBL00D1Absorbance, Fluorescence Intensity, LuminescenceTop and Bottom1SkanIt PC Software, Research Edition
VLBL0TD1Absorbance, Fluorescence Intensity, Luminescence, Time-resolved FluorescenceTop and Bottom1SkanIt PC Software, Research Edition
VLBL00D2Absorbance, Fluorescence Intensity, LuminescenceTop and Bottom2SkanIt PC Software, Research Edition
VLBL0TD2Absorbance, Fluorescence Intensity, Luminescence, Time-resolved FluorescenceTop and Bottom2SkanIt PC Software, Research Edition
VLBLATD2Absorbance, Fluorescence Intensity, Luminescence, Time-resolved Fluorescence, AlphaScreenTop and Bottom2SkanIt PC Software, Research Edition
VLBL0TD0Absorbance, Fluorescence Intensity, Luminescence, Time-resolved FluorescenceTop and BottomNoneSkanIt PC Software, Research Edition
Catalog number VLBLATD1
Price (INR)
-
Request A Quote
Measurement Technology:
Absorbance, Fluorescence Intensity, Luminescence, Time-resolved Fluorescence, AlphaScreen
Reading Options:
Top and Bottom
No. of Dispensers:
1
Includes:
SkanIt PC Software, Research Edition

Designed for bioscience researchers with a wide variety of needs, the Thermo Scientific™ Varioskan™ LUX multimode microplate reader comes equipped with a flexible range of measurement technologies including Absorbance, Fluorescence Intensity, Luminescence, AlphaScreen, and Time-Resolved Fluorescence. The instrument simplifies measurement setup with the automatic dynamic range selection, and its smart safety controls ease workflow and help avoid experimental errors. Varioskan LUX raises the bar for reliability and ease, with technology that works in researchers' best interest.

The Varioskan LUX multimode reader has received HTRF™ certification from Cisbio Bioassays and DLReady™ validation from Promega Corporation.

The Varioskan LUX is a modular, upgradable instrument that provides excellent usability across a variety of applications.

  • Up to five measurement technologies: Absorbance (UV-Vis), Fluorescence intensity (incl. FRET), Luminescence (direct and filtered), AlphaScreen/AlphaLISA and Time-resolved fluorescence (incl. TR-FRET/hTRF)
  • Filter and monochromator optics
  • Spectral scanning for assay optimization
  • Five measurement modes including end point, kinetic, spectra, kinetic spectra and multipoint
  • Integrated gas control module for setting CO2 and O2 concentrations for cell based assays
  • Simultaneous dispensing and measurement for signal monitoring at the start of a reaction
  • Automatic dynamic range selection, based on signal intensity of each well
  • Smart safety controls designed to identify and alert the user of potential problems before they happen
  • Automated instrument calibration and self-diagnostics for reliable performance
  • Certified HTRF compatible reader by CisBio Bioassays
  • Validated DLReady by Promega Corporation

Varioskan LUX is paired with the powerful and intuitive SkanIt Software, providing excellent usability and flexibility.

  • License free—Each user can install the software on his/her computer
  • Intuitive and flexible interface for multiple users and assays
  • Innovative Virtual Pipetting Tool makes it easy to define samples to plate layout
  • Measurement data is continuously saved to the database, so there's no data lost due to unexpected interruptions such as power outages or accidental aborting
  • Simplified data processing and export
    • Export data in a single mouse click, or have it emailed automatically at the end of the run
    • Several file formats for exporting results: *.xlsx, *.pdf, *.xml, and *.txt
  • Shows real-time spectra and kinetic curves during the measurement
  • Built-in calculations for data analysis (e.g. blank subtraction, average/SD/CV%, standard curve, dose response, quality control, classification, kinetic and spectral analysis, custom formula, etc.)
  • Comprehensive result reports with traceability to runtime actions and settings
  • Two editions available: Research Edition for life science researchers, and Drug Discovery Edition which offers features needed for the FDA 21 CFR Part 11 compliance.

The Varioskan LUX is modular and upgradeable, so you can build the system that best suits your needs. For more information, contact a sales representative.

Specifications
Accuracy0.003 Abs or ±2%, at 200 to 399 nm (0 to 2 Abs)
0.003 Abs or ±1%, at 400 to 1000 nm (0 to 3 Abs)
Accuracy (Photometric)0.003 Abs or ±2% at 200 to 399 nm (0 to 2 Abs), 0.003 Abs or ±1%, at 400 to 1000 nm (0 to 3 Abs)
CompatibilityRobot
Depth (English)23 in.
Depth (Metric)58 cm
Dimensions (L x W x H)58 x 53 x 51 cm (23 x 21 x 20 in.)
Dispense Volume2 – 5 000 μl with 1 μl increments
Dynamic RangeFluorescence intensity: top reading > 6 decades, bottom reading > 5.5 decades, Time-resolved fluorescence: > 6 decades, Luminescence: > 7 decades
Emission FiltersAvailable for purchase
Excitation FiltersFixed filters (Alpha Screen/AlphaLISA excitation wavelength 680 nm; TRF 334 nm)
Height (English)20 in.
Height (Metric)51 cm
IncludesSkanIt PC Software, Research Edition
Linearity96-well plate: ±2% (0 to 4 Abs at 450 nm), 384-well plate: ±2% (0 to 3 Abs at 450 nm)
Measurement Speed15 s for 96-well Plate, 45 s for 384-well Plate, 135 s for 1536-Well Plate (Minimum Times)
Measurement TypeEndpoint, Kinetic, Spectra, Multipoint, Kinetic Spectra
No. of Dispensers1
PrecisionSD < 0.001 Abs or CV < 0.5%, at 450 nm (0 to 3 Abs)
SensitivityFluorescence intensity (top reading): < 0.4 fmol fluorescein/well (black 384-well plate), Fluorescence intensity (bottom reading): < 4 fmol fluorescein/well, (clear bottom black 384 square well plate), Time-resolved fluorescence: < 1 amol/well (white low volume 384 well plate)Luminescence: < 7 amol ATP/well (white 384-well plate), AlphaScreen: < 100 amol phosphotyrosine/well (384-well plate)
Software Language VersionsEnglish, German, French, Spanish, Portuguese, Italian, Japansese, Chinese
Spectral Scanning Speed< 2.2 s/Well 400 to 500 nm, 1 Flash, 2 nm Step
Wavelength RangeAbsorbance: 200-1000 nm; Fluorescence intensity: Excitation: 200-822 nm, Emission: 270-840 nm: TRF: Excitation 334 nm, Emission 400 to 700 nm; Luminescence: 360-670 nm; AlphaScreen: Excitation 680 nm, Emission 400-660 nm
Weight (English)128 lb
Weight (Metric)58 kg
Width (English)21 in.
Width (Metric)53 cm
Detector TypePhotomultiplier Tube (PMT)
Incubator TemperatureAmbient +4° to 45°C
InterfacePC Software (Thermo Scientific SkanIt Software)
Light SourceXenon Flash Lamp, LED
Measurement TechnologyAbsorbance, Fluorescence Intensity, Luminescence, Time-resolved Fluorescence, AlphaScreen
Plate Type6 to 1536-well Plates (Absorbance 6 to 384-well Plates), μDrop Plates
Product LineVarioskan
Reading OptionsTop and Bottom
Read-out Range0 to 6 Abs
ShakingOrbital
Software TypeSkanIt Software
Wavelength SelectionDouble Monochromator (Absorbance and Spectral Scanning), Double Excitation and Emission Monochromator (Fluorescence Intensity), Filter (AlphaScreen and Time-Resolved Fluorescence)
Unit SizeEach

Frequently asked questions (FAQs)

In the Varioskan LUX Multimode Microplate Reader, how is running a TRF spectrum by setting excitation at 335 nm and using emission range different from running a filter-based TRF assay? Why does SkanIT Software have the TRF spectrum option?

The TRF spectrum option is useful for spectral analysis, assay and label development, and optimization purpose. For example, when a new TRF chelate label is developed, TRF spectrum measurement can be used to analyze the spectra when a lot of label is loaded in the well.

When the TRF spectrum option is used, the sensitivity is not sufficient for actual TRF assays as the instrument sensitivity decreases by about 200-fold. Hence, the TRF spectrum option should not be used for normal TRF assay applications. For optimal results, the TRF spectrum option should be used for TRF spectral scanning but not for running actual TRF assays. Actual TRF assays must be performed using filter-based TRF optics in the LAT module.

If the Varioskan unit does not have TRF optics in the LAT module, TRF spectral scanning cannot be performed. In this case, TRF spectral scanning can only be performed using TRF-based filters.

Find additional tips, troubleshooting help, and resources within our Cell Analysis Support Center.

How to remove a stuck microplate from the Varioskan LUX Multimode Microplate Reader?



Power off the Varioskan LUX instrument, open the sliding door, and then press both springs outward (1) to unlock the bottom of the front panel. Tilt the bottom of the panel outwards slightly and lift up to remove.



Look inside the instrument to locate the tray (3). Gently slide the tray to the left and then towards you to access the plate. Check for jams and make sure that the correct holder is installed. When finished, push the tray back into the instrument (exact position doesn’t matter as long as it is completely inside) and reinstall the front panel.

Note: Remove the bottom front cover on the instrument, in order to access the plate carrier, and manually pull out the plate. Do not attempt to grab the plate back through the plate door, otherwise it can cause alignment issues that will require a repair.

Find additional tips, troubleshooting help, and resources within our Cell Analysis Support Center.

Can relative fluorescence units (RFUs) be compared between two different plate readers?

Relative fluorescence units (RFUs) cannot be compared between two different microplate readers. RFU values are arbitrary and dimensionless, meaning that the actual numbers don't have a specific physical meaning, but the relative values are used for comparison. The critical measurement is the ratio of signal to background.

Whether the signal is low or high is 100% dependent on the baseline, i.e., if the sample signal is 2 and the baseline is 1, then it is a low signal but if the sample signal is 2 and the baseline is 0.01, then it is a very high signal. Instruments that typically give sample signals in the tens of thousands RFUs also typically have baseline RFUs somewhere in the tens or hundreds. For example, the Varioskan Microplate Reader is designed such that the baseline is kept below 0.1 RFU.

How can I move the Varioskan LUX Multimode Microplate Reader from one place to another?

We recommend transporting the Varioskan LUX Multimode Microplate Reader in the original packaging it was delivered in, with proper padding in the box, and after securing the tray holder transport lock on the instrument. See page 84 of the user manual for instructions to refit the transport lock of the tray holder.

Note: We also offer a service to move the instrument. Please reach out to serviceproducts@thermofisher.com.

Find additional tips, troubleshooting help, and resources within our Cell Analysis Support Center.

Which is the best DNA or RNA quantification instrument?

All of the DNA instruments offered by Thermo Fisher Scientific are of excellent quality. To find the right instrument to quantify DNA or RNA in your lab, explore the detailed comparison of UV-Vis spectrophotometers and fluorometers. Things like sensitivity, throughput and budget may be initial considerations when selecting a DNA quantification instrument. You may also consider whether you need target specificity, sample purity information, RNA quality information, or broad dynamic range.

Find additional tips, troubleshooting help, and resources within our Nucleic Acid Quantification Support Center.