Search
Search
| Catalog Number | Product Type |
|---|---|
| VLUPLAT08 | Upgrade kit for luminometry, AlphaScreen and TRF |
| N16443 | Storage Holder for Plate Adapters |
| N16484 | Emission Filter Pick-up Tool |
| VLUPA003 | AlphaScreen upgrade kit |
| VLUPD005 | Dispenser Upgrade Kit |
| VLUPT004 | TRF Upgrade Kit |
| N02693 | 96-well Adapter for Plate w/ Lid |
| N02696 | 6-48-well Adapter for Plate w/o Lid |
| N03079 | Robotic plate tray for plates with and without lids |
| N06210 | 1536-well Adapter for 10 mm Plate w/o Lid |
| SP-00094 | Dispensing tube assembly for a dispenser |
| SP-00096 | Dispenser syringe, 1 mL |
| N16308S | Liquid Protection Tray |
| VLUPLA06 | Upgrade kit for luminometry and AlphaScreen |
| VLUPLT07 | Upgrade Kit for Luminometry and TRF |
| VLUPD209 | Upgrade kit for two dispensers |
Thermo Scientific™ Varioskan™ LUX multimode microplate reader is a versatile tool for busy labs equipped with a flexible range of measurement technologies including Absorbance, Fluorescence Intensity, Luminescence, AlphaScreen, and Time-Resolved Fluorescence. You can configure the instrument to your needs, and when your research focus changes, just upgrade the instrument with service installable upgrade kits. Various plate adapters and other accessories helps you to optimize your assays.