Vanquish™ High-throughput LC systems
Actual product may vary
Vanquish™ High-throughput LC systems
Vanquish™ High-throughput LC systems
Thermo Scientific™
Vanquish™ High-throughput LC systems
Vanquish High-throughput LC systems can boost your lab's productivity by increasing throughput and reducing cost per sample without added bench space. Vanquish Duo UHPLC Systems are an ACT labeled product with a My Green Lab ACT Ecolabel.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberModelFor Use With (Application)
VQDUO-TANDEMVanquish Tandem LC SystemsIncrease lab throughput and return on investment for Tandem LC or LC-MS workflows. Provides offline column regeneration and efficiently uses your detector or mass spectrometer and maximizes the output. Setup methods with smart Chromeleon CDS wizards.
VQDUO-DUALLCVanquish Duo UHPLC SystemsTwo channel LC doubles throughput, deepens sample knowledge, saves on bench space, and improves return on investment. Control the system with easy to use Chromeleon CDS.
Catalog number VQDUO-TANDEM
Price (INR)
-
Model:
Vanquish Tandem LC Systems
For Use With (Application):
Increase lab throughput and return on investment for Tandem LC or LC-MS workflows. Provides offline column regeneration and efficiently uses your detector or mass spectrometer and maximizes the output. Setup methods with smart Chromeleon CDS wizards.
Use the Thermo Scientific™ Vanquish™ High-throughput LC Systems to improve productivity and save bench space. Vanquish Tandem LC systems increase sample throughput and MS utilization for LC-MS applications by eliminating column washing and equilibration steps from method cycle time. Vanquish Duo UHPLC systems combine two completely independent LC flow paths to double sample throughput or run independent analyses in parallel. In addition, Vanquish High-throughput LC Systems expand the benefits of the Vanquish platform without compromising performance, robustness, or ease-of-use.
Two flow paths. One integrated system.
  • Increase your sample throughput
  • Free up valuable bench space
  • Enhance qualitative and quantitative sample knowledge
  • Reduce your cost per sample
  • Streamline your workflow using intelligent software tools
Vanquish Duo UHPLC systems
Double your throughput, deepen your sample knowledge, and better utilize your bench space with the Vanquish Duo system, a two channel LC for improved return on investment.
Vanquish Tandem LC systems
Increase your lab's throughput and return on investment for LC-MS assays with the Vanquish Tandem LC system for offline column regeneration.
More productivity with ease
  • Save time spent on user training with Thermo Scientific™ Chromeleon™ Chromatography Data System (CDS) wizards for creating Vanquish High-throughput system methods.
  • Control two independent flow paths from a single user interface with fully compliant Chromeleon CDS.
  • Save time using the Chromeleon CDS software for data analysis.
  • Seamlessly integrate with a mass spectrometry or other detector.
Sustainability
The Vanquish Duo UHPLC Systems are an ACT labeled product with a My Green Lab ACT Ecolabel.
The My Green Lab ACT Ecolabel serves as the premier ecolabel for lab products, encompassing consumables, chemicals, and equipment. Standing for Accountability, Consistency, and Transparency (ACT), this certification offers clear, standardized information about a product's environmental footprint across its life cycle covering manufacturing, energy and water use, packaging, and end-of-life disposal. Leverage the My Green Lab ACT Ecolabel to make informed purchasing choices based on transparent environmental data so you can advance sustainability without sacrificing performance. Learn more about the ACT Ecolabel and participating Thermo Scientific products here.
Our general purpose product lines are not intended for in vitro diagnostic purposes in accordance with our product documentation, manuals, and labels. They are designated for General Laboratory Use Only.
Our general purpose product lines have not been tested or validated for such applications and their use for in vitro diagnostic purposes may result in health and safety risks.
For General Lab Use Only. Not For Diagnostic Procedures.
Specifications
DescriptionVanquish Tandem LC Systems
For Use With (Application)Increase lab throughput and return on investment for Tandem LC or LC-MS workflows. Provides offline column regeneration and efficiently uses your detector or mass spectrometer and maximizes the output. Setup methods with smart Chromeleon CDS wizards.
ModelVanquish Tandem LC Systems
For Use With (Equipment)Vanquish Horizon, Vanquish Flex or Vanquish Core Modules
Max. Pressure150 MPa, depending on configuration
TypeHPLC or Biocompatible UHPLC
Unit SizeEach