Wash buffer can be used for all ELISA assays.Read more
Have Questions?
Catalog Number
Quantity
WB01
100 mL
Catalog number WB01
Price (INR)
4,016.00
キャンペーン価格
Ends: 31-Dec-2026
6,693.00
Save 2,677.00 (40%)
100 ml
Quantity:
100 mL
Request bulk or custom format
Price (INR)
4,016.00
キャンペーン価格
Ends: 31-Dec-2026
6,693.00
Save 2,677.00 (40%)
100 ml
Wash buffer can be used for all ELISA assays.
Specifications
Shipping ConditionWet Ice
Quantity100 mL
Unit Size100 ml
Contents & Storage
Buffer. Store at 2°C to 8°C.
Citations & References (1)
Citations & References
Abstract
Novel method of monitoring trace cytokines and activated STAT molecules in the paws of arthritic mice using multiplex bead technology.
Authors:Lu LD, Stump KL, Seavey MM
Journal:BMC Immunol
PubMed ID:21073728
The use of mouse models to study human disease provides useful data that can provide support for research projects or an existing drug discovery program. How well a model recapitulates the human condition and the ease and reproducibility of data collected will determine how much confidence a scientist can place ... More