Product Image
LOWER BUFFER CHAMBER SPARE
Have Questions?
Catalog number WR0102
Price (INR)
97,923.00
1 each
Price (INR)
97,923.00
1 each
LOWER BUFFER CHAMBER SPARE
Specifications
Unit Size1 each
Frequently asked questions (FAQs)
Can I purchase replacement parts for the XCell4 SureLock Midi-Cell?
Here are the replacement parts we offer for the XCell4 SureLock Midi-Cell. They can only be ordered by phone:
- XCell4 SureLock Midi-Cell Lid: Cat. No. WR0101
- XCell4 SureLock Midi-Cell Lower Buffer Chamber: Cat. No. WR0102
- XCell4 SureLock Midi-Cell Displacement Dam: Cat. No. WR0106
- XCell SureLock Upgrade kit: Cat. No. WR0112 (contains 1 SureLock Assembly, 1 buffer dam, and 2 cores) Please note that in addition to ordering on the phone by calling Customer Service, this item can also be ordered using the “Quick Order” link (https://www.thermofisher.com/store/quick-order) that can be found at the top right of most pages on our website. The price will display when the item is added to your shopping cart.
Find additional tips, troubleshooting help, and resources within our Protein Gel Electrophoresis Chambers, Power Supplies, and Accessories Support Center.
What are the components of the XCell4 SureLock Midi-Cell?
The XCell4 SureLock Midi-Cell includes the lower buffer chamber, lid with cables, two buffer cores, buffer dam, displacement dam, SureLock assembly, and gel knife. For descriptions and images of the various parts, please refer to pages 1-3 of the product manual (https://assets.thermofisher.com/TFS-Assets/LSG/manuals/xcell4_midicell_man.pdf).
Find additional tips, troubleshooting help, and resources within our Protein Electrophoresis and Western Blotting Support Center.