Sterilin™ Water Sampling Bottles
Actual product may vary
Sterilin™ Water Sampling Bottles
Sterilin™ Water Sampling Bottles
Thermo Scientific™
Sterilin™ Water Sampling Bottles
Conveniently sample both chlorinated or nonchlorinated water for microbial analysis with Thermo Scientific™ Sterilin™ Water Sampling Bottles.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacity (Metric)ShapeDescriptionMaterial
WSC1000-201000 mLSquareDosed 20mg/LPET
WSC350NT
also known as WSC350/NT
350 mLRectangularUndosedPET
WSC350350 mLRectangularDosed 120mg/LPET
WSC500500 mLSquareDosed 120mg/LPET
WSC500-20500 mLSquareDosed 20mg/LPET
WSC500NT
also known as WSC500/NT
500 mLSquareUndosedPET
WSC500T500 mLRectangularDosed 120mg/LPET
WSC500T-20500 mLRectangularDosed 20mg/LPET
WSC500TNT
also known as WSC500T/NT
500 mLRectangularUndosedPET
500WSC500 mLRoundDosed 20mg/LPolystyrene
WSC10001000 mLSquareDosed 120mg/LPET
WSC1000NT
also known as WSC1000/NT
1000 mLSquareUndosedPET
Catalog number WSC1000-20
Price (INR)
-
Capacity (Metric):
1000 mL
Shape:
Square
Description:
Dosed 20mg/L
Material:
PET
Conveniently sample both chlorinated or nonchlorinated water for microbial analysis with Thermo Scientific™ Sterilin™ Water Sampling Bottles.
Not available in all geographic regions.
Features:
  • Tamper evident for sample integrity
  • Lot number for complete traceability
  • Sterilized by gamma irradiation
  • Available empty, or pre-dosed with sodium thiosulphate
Available Manufactured Using:
  • Flexible polystyrene
  • NEW, lightweight PET for improved handling
Polystyrene Bottles Feature Color-Coded Labels to Aid Differentiation:
  • Blue: dosed
  • Green: undosed
Available Dose Levels:
  • 20mg/h: Suitable for neutralizing water samples with a low concentration of chlorine
  • 120mg/h: Suitable for neutralizing water samples with a high concentration of chlorine
Specifications
Closure MaterialHDPE
DescriptionDosed 20mg/L
No. per Case84
ShapeSquare
Capacity (Metric)1000 mL
ColorNatural
MaterialPET
SterilityGamma-irradiated
Unit SizeEach