NuPAGE™ Tris-Acetate Midi Protein Gels, 3 to 8%, 1.0 mm, WedgeWell™ format
NuPAGE™ Tris-Acetate Midi Protein Gels, 3 to 8%, 1.0 mm, WedgeWell™ format
NuPAGE™ Tris-Acetate Midi Protein Gels, 3 to 8%, 1.0 mm, WedgeWell™ format
NuPAGE™ Tris-Acetate Midi Protein Gels, 3 to 8%, 1.0 mm, WedgeWell™ format
Invitrogen™

NuPAGE™ Tris-Acetate Midi Protein Gels, 3 to 8%, 1.0 mm, WedgeWell™ format

Invitrogen NuPAGE Tris-Acetate protien gels, WedgeWell format provide excellent separation of large molecular weight proteins. They are high performance polyacrylamide gels that simulate the denaturing or native conditions of the traditional Laemmli system.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberWells
WTA03826BOX26-well
WTA03812BOX12 + 2-well
WTA03820BOX20-well
Catalog number WTA03826BOX
Price (INR)
14,201.00
Online offer
Ends: 31-Mar-2026
23,667.00
Save 9,466.00 (40%)
10 gels (1 box)
Add to cart
Wells:
26-well
Price (INR)
14,201.00
Online offer
Ends: 31-Mar-2026
23,667.00
Save 9,466.00 (40%)
10 gels (1 box)
Add to cart
Invitrogen NuPAGE Tris-Acetate protein gels, WedgeWell format provide excellent separation of large molecular weight proteins. They are high performance polyacrylamide gels that simulate the denaturing or native conditions of the traditional Laemmli system. The unique buffer formulation maintains a low operating pH during electrophoresis, resulting in superior resolution of high molecular weight proteins compared to traditional Tris-glycine SDS-PAGE gels.

Invitrogen NuPAGE Tris-Acetate WedgeWell format protein gels provide excellent separation of large molecular weight proteins. They are high performance polyacrylamide gels that simulate the denaturing or native conditions of the traditional Laemmli system. The unique buffer formulation maintains a low operating pH during electrophoresis, resulting in superior resolution of high molecular weight proteins compared to traditional Tris-glycine SDS-PAGE gels.

Product features

  • High resolution—gels offer optimal separation of high molecular weight proteins
  • Better protein integrity—sample preparation process has been optimized to help preserve proteins
  • Longer shelf life—gels can be stored for nine months at room temperature
  • Wedge-shaped wells—for easier sample loading and up to two times higher sample volume

Run your proteins in native or denatured form

NuPAGE Tris-Acetate protein gels do not contain SDS and can be used to separate proteins in native or denatured form. For denatured proteins, we recommend using NuPAGE LDS Sample Buffer (NP0007) and NuPAGE Tris-Acetate SDS Running Buffer (LA0041). For native proteins, we recommend using Novex Tris-Glycine Native Sample Buffer (LC2673) and Novex Tris-Glycine Native Running Buffer (LC2672).

NuPAGE WedgeWell Tris-Acetate Midi gels can be run in the SureLock Tandem Midi Gel Tank (STM1001) or XCell4 SureLock Midi-Cell (WR0100).

For transfer of proteins to a membrane, rapid semi-dry transfer can be done using the Power Blotter or rapid dry transfer using the iBlot 3 Gel Transfer Device (IB31001). Alternatively, traditional wet transfer can be performed using the SureLock Tandem Midi Blot Module (STM2001) using NuPAGE Transfer buffer (NP00061).

Convenient to use

NuPAGE Tris-Acetate gels, WedgeWell format, have innovative wedge-shaped wells that allow up to two times more sample load volume than other 1.0 mm-thick gels. The wedge-shaped wells also have larger openings, making sample loading easier.

For Research Use Only. Not for use in diagnostic procedures.
Specifications
For Use With (Equipment)XCell4 SureLock Midi-Cell
Gel Thickness1.0 mm
Length (Metric)8 cm
Mode of SeparationMolecular Weight
Quantity10 gels (1 box)
Sample Loading VolumeUp to 40 μL
Shelf Life274 Days
Shipping ConditionWet Ice
Width (Metric)13 cm
Gel Percentage3 to 8%
Gel SizeMidi
Gel TypeTris-Acetate
Product LineNuPAGE
Separation Range36 to 500 kDa
Separation TypeDenaturing, Native
Wells26-well
Unit Size10 gels (1 box)
Contents & Storage
Store at 4°C to 25°C. Do not freeze.

Frequently asked questions (FAQs)

What are the recommended maximum sample loading volumes for the new WedgeWell format holds for the midi gels?

For the 12+2 well large gels, the maximum sample loading volume is 100 µL. For the 12+2 well small gels, the maximum sample loading volume is 90 µL. For 20 well gels, the maximum sample loading volume is 60 µL and for the 26 well gels, the maximum sample loading volume is 40 µL.

Find additional tips, troubleshooting help, and resources within our Protein Gel 1D Electrophoresis Support Center.