Novex™ Tris-Glycine Plus Midi Protein Gels, 4 to 20%, 1.0 mm, WedgeWell™ format
Novex™ Tris-Glycine Plus Midi Protein Gels, 4 to 20%, 1.0 mm, WedgeWell™ format
Novex™ Tris-Glycine Plus Midi Protein Gels, 4 to 20%, 1.0 mm, WedgeWell™ format
Novex™ Tris-Glycine Plus Midi Protein Gels, 4 to 20%, 1.0 mm, WedgeWell™ format
Invitrogen™

Novex™ Tris-Glycine Plus Midi Protein Gels, 4 to 20%, 1.0 mm, WedgeWell™ format

Novex Tris-Glycine Plus midi protein gels, WedgeWell format are improved polyacrylamide gels based on the traditional Laemmli protein chemistry, allowing the use of Laemmli sample and running buffers. The gels offer reproducible separation of a wide range of proteins into well-resolved bands.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberWells
WTG42026BOX26-well
WTG42012BOX12 + 2-well
WTG42020BOX20-well
Catalog number WTG42026BOX
Price (INR)
17,055.00
Online offer
Ends: 31-Mar-2026
24,364.00
Save 7,309.00 (30%)
10 gels (1 box)
Add to cart
Wells:
26-well
Price (INR)
17,055.00
Online offer
Ends: 31-Mar-2026
24,364.00
Save 7,309.00 (30%)
10 gels (1 box)
Add to cart
Novex Tris-Glycine Plus midi protein gels, WedgeWell format are improved polyacrylamide gels based on the traditional Laemmli protein chemistry, allowing the use of Laemmli sample and running buffers. The gels offer reproducible separation of a wide range of proteins into well-resolved bands. Novex Tris-Glycine Plus WedgeWell format midi gels are a 1.0-mm thick, wider format (8 x 13 cm) gel for higher throughput electrophoresis of protein samples.

Novex Tris-Glycine Plus WedgeWell format midi protein gels are improved polyacrylamide gels based on the traditional Laemmli protein chemistry, allowing the use of Laemmli sample and running buffers. The gels offer reproducible separation of a wide range of proteins into well-resolved bands. Novex Tris-Glycine Plus WedgeWell format midi gels are a 1.0-mm thick, wider format (8 x 13 cm) gel for higher throughput electrophoresis of protein samples.

Product features

  • Flexibility—use for native and denaturing protein assays
  • Fast run conditions—quickly separate your proteins using constant voltage in less than 60 minutes
  • Wedge-shaped wells—for easier sample loading and up to two times higher sample volume

Run your proteins in native or denatured form

Novex Tris-Glycine Plus gels do not contain SDS and so can be used to separate proteins in a native or denatured state. For denatured proteins, we recommend using Novex Tris-Glycine SDS Sample Buffer (LC2676) and Novex Tris-Glycine SDS Running Buffer (LC26754). For native proteins, we recommend using Novex Tris-Glycine Native Sample Buffer (LC2673) and Novex Tris-Glycine Native Running Buffer (LC2672). The gels can be run in the SureLock Tandem Midi Gel Tank (STM1001) or XCell4 SureLock Midi-Cell (WR0100).

For transfer of proteins to a membrane, we recommend using the Novex Tris-Glycine Transfer Buffer (LC3675) when performing traditional wet transfer with the SureLock Tandem Midi Blot Module (STM2001). Rapid semi-dry transfer can be done using the Invitrogen Power Blotter or rapid dry transfer using the iBlot 3 Gel Transfer Device (IB31001).

Convenient to use

Novex Tris-Glycine Plus WedgeWell format gels have innovative wedge-shaped wells that allow up to two times more sample load volume than other 1.0 mm-thick gels. The wedge-shaped wells also have larger openings, which makes sample loading easier.

For Research Use Only. Not for use in diagnostic procedures.
Specifications
For Use With (Equipment)SureLock Tandem Midi Gel Tank
Gel Thickness1.0 mm
Length (Metric)8 cm
Mode of SeparationMolecular Weight
Quantity10 gels (1 box)
Sample Loading VolumeUp to 40 μL
Shelf Life180 Days
Shipping ConditionWet Ice
Width (Metric)13 cm
Gel Percentage4 to 20%
Gel SizeMidi
Gel TypeTris-Glycine Plus
Product LineNovex
Separation Range20 to 200 kDa
Separation TypeDenaturing, Native
Wells26-well
Unit Size10 gels (1 box)
Contents & Storage
Store at 2°C to 8°C. Do not freeze.

Frequently asked questions (FAQs)

Can I prepare my protein sample with the reducing agent and store it for future use?

DTT is not stable, so it must be added and the reduction performed just prior to loading your samples.

Find additional tips, troubleshooting help, and resources within our Protein Gel 1D Electrophoresis Support Center.

My LDS or SDS sample buffer precipitates when stored at 4 degrees C. Can I warm it up? Can I store it at room temperature?

Precipitation of the LDS or SDS at 4 degrees C is normal. Bring the buffer to room temperature and mix until the LDS/SDS goes into solution. If you do not want to wait for it to dissolve, you can store the sample buffer at room temperature.

Find additional tips, troubleshooting help, and resources within our Protein Gel 1D Electrophoresis Support Center.

How are Bolt gels different than NuPAGE gels?

While they are both Bis-Tris based gels, the chemistries are very different since Bolt gels are optimized for western blotting. Another key difference is the wedge well design of the Bolt gels, which allows larger sample volumes to be loaded.

Find additional tips, troubleshooting help, and resources within our Protein Gel 1D Electrophoresis Support Center.

Do I need to add water or buffer to the Displacement Dam when I run one or two gels in the XCell4 SureLock Midi-Cell?

The XCell4 Displacement Dam is used when fewer than 3 gels are run in the XCell4 SureLock Midi-Cell. The Displacement Dam is placed in the lower buffer chamber instead of a second buffer core. Do not add any buffer or water to the Displacement Dam. However, if water or buffer is accidentally added to the Displacement Dam, it does not affect the electrophoresis run.

Find additional tips, troubleshooting help, and resources within our Protein Electrophoresis and Western Blotting Support Center.

What are the maximum load volumes for the NuPAGE Midi Gels?

Recommended maximum load volumes:
12 + 2-well gel: 45 µL in sample well, 15 µL in marker well
20-well gel: 25 µL
26-well gel: 15 µL

Find additional tips, troubleshooting help, and resources within our Protein Electrophoresis and Western Blotting Support Center.