WedgeWell™ Empty Gel Cassettes with Combs, midi
Invitrogen™

WedgeWell™ Empty Gel Cassettes with Combs, midi

These WedgeWell empty gel cassettes are pre-assembled, single-use, plastic cassettes with combs that allow you to pour your own midi-size gels. The gels can be run in the SureLock Tandem Midi Gel Tank (STM1001) or XCell4 SureLock Midi-Cell (WR0100).
Have Questions?
Change viewbuttonViewtableView
Catalog NumberNo. of Wells
WW101212 + 2-well
WW102020-well
WW102626-well
Catalog number WW1012
Price (INR)
8,694.00
キャンペーン価格
Ends: 30-Sep-2026
11,592.00
Save 2,898.00 (25%)
1 pack
No. of Wells:
12 + 2-well
Price (INR)
8,694.00
キャンペーン価格
Ends: 30-Sep-2026
11,592.00
Save 2,898.00 (25%)
1 pack
These WedgeWell empty gel cassettes are pre-assembled, single-use, plastic cassettes with combs that allow you to pour your own midi-size gels. The gels can be run in the SureLock Tandem Midi Gel Tank (STM1001) or XCell4 SureLock Midi-Cell (WR0100). The cassettes are sealed on three sides to prevent leaking and do not require the use of a special casting stand. Gels poured in these cassettes are 1.0-mm thick. SureCast reagents can used to make pouring your own high performance polyacrylamide midi gels both easy and reproducible.

These WedgeWell empty gel cassettes are pre-assembled, single-use, plastic cassettes with combs that allow you to pour your own midi-size gels. The gels can be run in the SureLock Tandem Midi Gel Tank (STM1001) or XCell4 SureLock Midi-Cell (WR0100). The cassettes are sealed on three sides to prevent leaking and do not require the use of a special casting stand. Gels poured in these cassettes are 1.0-mm thick. SureCast reagents can used to make pouring your own high performance polyacrylamide midi gels both easy and reproducible.

Product features

  • The cassettes are sealed on three sides to prevent leaking and do not require the use of a special casting stand
  • Gels poured in these cassettes are 1.0-mm thick
  • SureCast reagents can be used to pour high-performance polyacrylamide midi gels

Handcast gel reagents

SureCast protein gels are a reliable and high-performance solution for your handcast gel electrophoresis needs. SureCast resolving and stacking buffers are based on the traditional Laemmli protein chemistry, allowing the use of Laemmli sample and running buffers. Handcast SureCast gels are a reliable, high-performance, low-cost option for gel electrophoresis.

Convenient to use

The midi WedgeWell empty gel cassettes have innovative wedge-shaped wells that allow up to two times more sample load volume than other 1.0 mm-thick gels. The wedge-shaped wells also have larger openings, which makes sample loading easier.

For Research Use Only. Not for use in diagnostic procedures.
Specifications
Length (Metric) Gel8 cm
No. of Wells12 + 2-well
Quantity1 pack
Running DimensionVertical
Shipping ConditionRoom Temperature
Thickness (Metric)1.0 mm
Width (Metric)13 cm
For Use With (Equipment)XCell4 SureLock Midi-Cell
Gel SizeMidi
TypeEmpty Gel Cassette
Unit Size1 pack
Contents & Storage
10 cassettes (with 12+2 well combs) per pack

Frequently asked questions (FAQs)

What are the recommended maximum sample loading volumes for the new WedgeWell format holds for the midi gels?

For the 12+2 well large gels, the maximum sample loading volume is 100 µL. For the 12+2 well small gels, the maximum sample loading volume is 90 µL. For 20 well gels, the maximum sample loading volume is 60 µL and for the 26 well gels, the maximum sample loading volume is 40 µL.

Find additional tips, troubleshooting help, and resources within our Protein Gel 1D Electrophoresis Support Center.

Do you offer empty cassettes and combs for casting Mini and Midi protein gels?

We do offer disposable, empty plastic cassettes and combs to provide the option of pouring your own Mini or Midi gels for use with the XCell SureLock Mini-Cell, Mini Gel Tank, or XCell4 SureLock Midi-Cell, respectively. All cassettes are sealed on three sides to prevent leaking. Since the cassettes are pre-sealed with the slot pre-taped, a bulky casting stand is not necessary. The empty cassettes and combs can be found here (https://www.thermofisher.com/us/en/home/life-science/protein-biology/protein-gel-electrophoresis/power-supplies-protein-electrophoresis.html#staining).

- The Empty Mini Gel Cassettes are available in two thicknesses (1.0 mm and 1.5 mm) and can be used with Invitrogen Combs that are available in two thicknesses (1.0 mm and 1.5 mm), and in 7 types (1 well, 2 well, 5 well, 10 well, 12 well, 15 well, and 2D well).
- The Bolt Empty Mini Gel Cassettes are available in 1.0 mm thickness and can be used with Bolt 10- or 12-well combs.
- The Empty Midi Gel Cassettes are available in 1.0 mm thickness and can be used with three Invitrogen Comb types (12 + 2 wells, 20 wells, and 26 wells).

Find additional tips, troubleshooting help, and resources within our Protein Electrophoresis and Western Blotting Support Center.