Attune™ Xenith™ Quality Control Beads
Invitrogen™
Attune™ Xenith™ Quality Control Beads
The Attune Xenith Quality Control Beads are designed for use with the Attune Xenith Flow Cytometer to characterize the instrument and monitor day-to-day performance.
Have Questions?
Catalog NumberQuantity
X200003 mL
Catalog number X20000
Price (INR)
26,082.00
25 tests
Quantity:
3 mL
Price (INR)
26,082.00
25 tests
Attune Xenith Quality Control Beads are for use with the Attune Xenith Flow Cytometer and its software to automatically characterize the instrument and monitor day-to-day instrument performance. Regular quality control (QC) of your instrument with these beads ensures the accuracy and sensitivity of the instrument and allows identification of any shifts in instrument performance.
The QC beads contain distinct sub-populations of beads labelled with different intensities of fluorescent signal and are used to establish optimal voltage settings for performing spectral and compensation experiments. The beads are pre-calibrated at the factory, and the lot-specific bead performance data is captured as a downloadable file for use in the Attune Xenith software. Each vial contains enough beads for 25 measurements.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Calibration TypeFlow Cytometry Quality Control
ColorFull spectral
FormatBeads in solution
No. of Tests25 Tests
Product LineAttune
Quantity3 mL
For Use With (Equipment)Attune Xenith Flow Cytometer
Product TypeTracking Beads
Unit Size25 tests
Contents & Storage
2°C to 8°C