Attune™ Xenith™ 50-nm Filter Kit
Invitrogen™
Attune™ Xenith™ 50-nm Filter Kit
The Attune Xenith 50-nm Filter Kit is designed for use with the Attune Xenith Flow Cytometer. This kit comprises one 50-nm primary focusing fluid filter and one set of filter caps that can be used to reduce background on the system caused by debris and particulates, enabling analysis of particles of 160 nm by scatter alone.
Have Questions?
Catalog NumberQuantity
X200501 Ea.
Catalog number X20050
Price (INR)
31,451.00
線上優惠
Ends: 30-Sep-2026
41,934.00
Save 10,483.00 (25%)
Each
Quantity:
1 Ea.
Price (INR)
31,451.00
線上優惠
Ends: 30-Sep-2026
41,934.00
Save 10,483.00 (25%)
Each
The Attune Xenith 50-nm Filter Kit is designed for use with the Attune Xenith Flow Cytometer. This kit contains one 50-nm primary focusing fluid filter and one set of filter caps. This filter can be used instead of the standard 200-nm primary fluid filter provided with the Attune Xenith Flow Cytometer to filter particulates larger than 50 nm in size out of the focusing fluid used to run the instrument as it passes into the system. This further reduces background on the system that can be caused by debris and particulates and enables analysis of smaller particles of 160 nm by scatter alone. The caps provided can be used to keep the filter wet when not in use on the instrument.
  • Filters out particulates >50 nm in size
  • Reduced background in the system caused by debris and particulates
  • Includes filter caps that can be used to keep the filter wet during storage if swapping between standard 200 nm filter and this 50 nm filter
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Quantity1 Ea.
For Use With (Equipment)Attune Xenith Flow Cytometer
Product TypeFilter
Unit SizeEach