Tetra Core Set and Novex™ Tris-Glycine 4-20% Gels Combos
Invitrogen™

Tetra Core Set and Novex™ Tris-Glycine 4-20% Gels Combos

The Tetra Electrophoresis Core Set and Novex Tris-Glycine 4-20% Combo includes the Tetra Electrophoresis primary and companion cores along with three boxes of Novex Tris-Glycine gels. Achieve broad-range protein separation with Laemmli-like chemistry in Bio-Rad's Mini-PROTEAN™ Tetra Cell tank, running up to four gels per run.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberWells
XP0420TS-12W12-well
XP0420TS-10W10-well
XP0420TS-15W15-well
Catalog number XP0420TS-12W
Price (INR)
26,315.00
Online offer
Ends: 31-Mar-2026
43,857.00
Save 17,542.00 (40%)
Each
Wells:
12-well
Price (INR)
26,315.00
Online offer
Ends: 31-Mar-2026
43,857.00
Save 17,542.00 (40%)
Each

The Invitrogen Tetra Electrophoresis Core Set and Novex Tris-Glycine 4-20% Combo provides a comprehensive solution for running high-performance Invitrogen precast protein gels in Bio-Rad's Mini-PROTEAN Tetra Cell. This combo product includes the Tetra Electrophoresis primary and companion cores along with three boxes of Novex Tris-Glycine gels, allowing for seamless integration and efficient protein analysis applications such as western blotting and SDS-PAGE. The intuitive design ensures ease of use, making it accessible for both new and experienced users.

Contents of the combo kit

  • Tetra Electrophoresis Primary Core (Cat. No. 1658037)
  • Tetra Electrophoresis Companion Core (Cat. No. 1658038)
  • 3 boxes of Novex Tris-Glycine 4-20% gels (10 gels per box)

About the Tetra Electrophoresis Core Set

The Tetra cores stand out due to their compatibility with all Invitrogen mini gels, allowing users to swap and run gels effortlessly while achieving high-performance results. When using both cores, up to four gels can be run simultaneously, which significantly increases throughput and efficiency. Additionally, the self-aligning cassettes and leak-proof design ensure stable and reliable electrophoresis runs, making experiment setup easy and fast.

The Tetra Electrophoresis Primary Core comes with the Tetra Buffer Dam (Cat. No. 1658039) for use when running only one gel in the core. To run three or four gels at once, the Tetra Electrophoresis Companion Core is used alongside the Primary Core. This combination allows for the simultaneous running of multiple gels, further enhancing throughput and efficiency in your lab. The Companion Core works seamlessly with the Primary Core, providing a flexible solution for varying experimental needs.

This cost-effective solution eliminates the need for new electrophoresis equipment, saving costs while enhancing your lab's capabilities. The Tetra Electrophoresis Core Set is straightforward to use: simply swap the cores in your Bio-Rad Mini-PROTEAN Tetra system for the Invitrogen Tetra cores and use Invitrogen mini protein gels following their recommended protocols.

About Novex Tris-Glycine gels, WedgeWell format

Novex Tris-Glycine mini gels are polyacrylamide gels based on traditional Laemmli protein electrophoresis, allowing the use of Laemmli sample and running buffers. They enable consistent performance and separation of a wide range of proteins into well-resolved bands. Novex Tris-Glycine gels, WedgeWell format, have innovative wedge-shaped wells that allow up to two times more sample load volume than other 1.0 mm-thick gels. The wedge-shaped wells also have larger openings, making sample loading easier.

For transferring your proteins to a membrane, we recommend using for Novex Tris-Glycine Native Running Buffer (Cat. No. LC2672) or Novex Tris-Glycine Transfer Buffer (Cat. No. LC3675) for traditional wet transfer using the XCell II Blot Module (Cat. No. EI9051). Alternatively, rapid semi-dry transfer can be performed using the Power Blotter or rapid dry transfer using the iBlot 3 Gel Transfer Device (Cat. No. IB31001).

For Research Use Only.
Specifications
AdaptersTetra Electrophoresis Cores
BufferNovex Tris-Glycine SDS Running Buffer, Novex Tris-Glycine Native Running Buffer
For Use With (Equipment)XCell SureLock Mini-Cell
Gel Thickness1.0 mm
Length (Metric)8 cm
Mode of SeparationMolecular Weight
QuantityTetra Core Set, 3 boxes of gels (30 gels)
Sample Loading VolumeUp to 45 μL
Shelf Life7 Months
Shipping ConditionWet Ice
Width (Metric)8 cm
Gel Percentage4 to 20%
Gel SizeMini
Gel TypeTris-Glycine Plus
Product LineNovex, WedgeWell
Product TypeElectrophoresis Cores and Precast Protein Gels
Separation Range20 to 200 kDa
Separation TypeDenaturing, Native
Separation ofProtein
Wells12-well
Unit SizeEach
Contents & Storage
Store at 2°C to 8°C. Do not freeze.