Chloroform (Approx. 0.75% Ethanol as Preservative/Molecular Biology), Fisher BioReagents
Actual product may vary
Chloroform (Approx. 0.75% Ethanol as Preservative/Molecular Biology), Fisher BioReagents
Chloroform (Approx. 0.75% Ethanol as Preservative/Molecular Biology), Fisher BioReagents
Fisher Bioreagents

Chloroform (Approx. 0.75% Ethanol as Preservative/Molecular Biology), Fisher BioReagents

CAS: 67-66-3 | CHCl3 | 119.37 g/mol
Have Questions?
Catalog NumberPackagingQuantity
YBP11451Glass Bottle1 L
Catalog number YBP11451
HSN 29031300
Price (INR)
30,524.00
Each
Packaging:
Glass Bottle
Quantity:
1 L
Glass bottle; 1L
This solvent is used in Phenol/Chloroform extractions to remove proteins from DNA or RNA samples.
Specifications
Chemical Name or MaterialChloroform
Cleanup SymbolSolvent
Melting Point-63°C
Name NoteApprox. 0.75% Ethanol as Preservative
Absorbance0.13 max. at 254nm, 0.28 max. at 250nm, 0.8 max. at 245nm, 1 max. at 244nm
CAS67-66-3
ChemAlert Storage SymbolBlue
ColorColorless
CAS64-17-5
Decomposition Information290°C
Recommended StorageRT
Density1.480g/cm³
Quantity1 L
PackagingGlass Bottle
Physical FormLiquid
Refractive Index1.4420 to 1.4450 (at 25°C)
Assay Percent Range≥99 % (by GC)
Residue after Evaporation2ppm max.
Molecular FormulaCHCl3
MDL NumberMFCD00000826
Vapor Pressure213mbar at 20°C
Viscosity0.56 mPa/s at 20°C
Water0.02% max.
Synonymtrichloromethane, formyl trichloride, methane, trichloro, trichloroform, methane trichloride, methenyl trichloride, methyl trichloride, trichlormethan, chloroforme, cloroformio
Boiling Point61°C
InChI KeyHEDRZPFGACZZDS-UHFFFAOYSA-N
SMILESClC(Cl)Cl
IUPAC Namechloroform
Molecular Weight (g/mol)119.37
PubChem CID6212
ChEBICHEBI:35255
Formula Weight119.38
Percent Purity≥99%
GradeMolecular Biology
Unit SizeEach