You need to enable JavaScript to run this app.
Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture and Transfection
Chemicals
Chromatography
Electron Microscopes
Lab Plasticware and Supplies
Lab Centrifuges
Lab Solutions
Mass Spectrometers
Next Generation Sequencers
See all product categories
Applications
Brands
Industrial and Applied Sciences
Food and Beverage
Forensics
Lab Solutions
Life Sciences
Pharma and Biopharma
Biotechnology
Clinical and Diagnostics
Digital Solutions
See all applications
Services
Lab Instrument and Equipment Services
Custom Services
Training Services
Financial and Leasing Services
Enterprise Level Lab Informatics
Partnering and Licensing Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Cell Biology Services
Food Safety Inspection Services
See all services
Help and Support
Contact Us
Product Documentation
Knowledge Base and Product FAQs
Learning Centers
Supply Center
eProcurement Solutions
Lab Instrument and Equipment Support
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Special Offers
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
Instrument Management
Home
▸
Shop All Products
▸
Facility Safety and Maintenance
▸
Cleaning Supplies and Equipment
▸
Cleaners and Disinfectants
▸
Laboratory Surfactants and Wetting Agents
▸
Sodium Dodecyl Sulfate (SDS), White Powder, Electrophoresis, Fisher BioReagents™
Element Search
Structure Search
Fisher Bioreagents
Sodium Dodecyl Sulfate (SDS), White Powder, Electrophoresis, Fisher BioReagents™
For protein extraction procedures and SDS polyacrylamide gels | CAS: 151-21-3 | C12H25NaO4S | 288.38 g/mol
Have Questions?
Change view
Quantity:
100 g
500 g
Packaging:
View all
Glass Bottle
Catalog number
YBP166100
HSN
29209010
Price (INR)
6,646.00
Each
Add to cart
Quantity:
100 g
Packaging:
Glass Bottle
Request bulk or custom format
Price (INR)
6,646.00
Each
Add to cart
Specifications
Absorbance
0.1 max. (0.1% solution) at 280nm, 0.4 max. (0.1% solution) at 230nm
Chemical Name or Material
Sodium Dodecyl Sulfate
Color
White
CAS
151-21-3
ChemAlert Storage Symbol
Red
View more
For protein extraction procedures and SDS polyacrylamide gels
DNase and RNase-free detergent for protein extraction procedures and polyacrylamide gel electrophoresis
SDS is a commonly used detergent in protein purification and polyacrylamide gel electrophoresis (PAGE)
SDS denatures proteins by disrupting the non-covalent bonds of the protein
Functionally tested for use in electrophoresis
Intended for research use only; not for human or animal diagnostic purposes