Forskolin (From Coleus Forskohlii), Fisher BioReagents
Forskolin (From Coleus Forskohlii), Fisher BioReagents
Forskolin (From Coleus Forskohlii), Fisher BioReagents
Fisher Bioreagents

Forskolin (From Coleus Forskohlii), Fisher BioReagents

Have Questions?
Change viewbuttonViewtableView
Quantity:
10 mg
Packaging:
Amber Glass
Catalog number YBP252010
HSN 29321990
Price (INR)
42,847.00
Each
Quantity:
10 mg
Packaging:
Amber Glass
Request bulk or custom format
Price (INR)
42,847.00
Each
Specifications
Chemical Name or MaterialForskolin
Name NoteFrom Coleus forskohlii
CAS66575-29-9
ChemAlert Storage SymbolGray
Melting Point228°C
View more
Forskolin is a molecular biology reagent. Forskolin functions as an antihypertensive and vasodilator. Adenylcyclase activator.