Sodium Tartrate Dihydrate (Colorless Crystals), Fisher BioReagents
Actual product may vary
Sodium Tartrate Dihydrate (Colorless Crystals), Fisher BioReagents
Sodium Tartrate Dihydrate (Colorless Crystals), Fisher BioReagents
Fisher Bioreagents

Sodium Tartrate Dihydrate (Colorless Crystals), Fisher BioReagents

CAS: 6106-24-7 | C4H8Na2O8 | 230.08 g/mol
Have Questions?
Catalog NumberQuantityPackaging
YBP352500500 gGlass Bottle
Catalog number YBP352500
HSN 28181000
Price (INR)
18,326.00
Each
Quantity:
500 g
Packaging:
Glass Bottle
Price (INR)
18,326.00
Each
Glass Bottle; 500g
This sodium salt is used in buffers for molecular biology and cell culture applications.
Specifications
Chemical Name or MaterialSodium Tartrate Dihydrate
CAS6106-24-7
ChemAlert Storage SymbolGray
Health Hazard 1SIGNAL WORD: CAUTION!
CAS868-18-8
Health Hazard 3Emergency Overview
May cause skin, eye, and respiratory tract irritation.
NFPA
Health: 1
Flammability: 1
Instability: 0
ColorWhite
Recommended StorageRT
pH7 to 9
Loss on Drying15.61 to 15.71% (at 150°C)
PackagingGlass Bottle
Molecular FormulaC4H8Na2O8
MDL NumberMFCD00150035
Quantity500 g
Merck Index15, 8812
Synonymunii-dia7c37aow, disodium tartrate dihydrate, disodium l-+-tartrate dihydrate, dia7c37aow, disodium +-tartrate dihydrate, sodium l-+-tartrate dihydrate, butanedioic acid, 2,3-dihydroxy-2r,3r-, disodium salt, dihydrate, disodium tartarate dihydrate, sodium tartrate nf, sodium tartrate dihydrate mi
Solubility Information≤0.005% Insoluble Matter
InChI KeyFGJLAJMGHXGFDE-UDMGOBQRNA-L
SMILESO.O.[Na+].[Na+].O[C@H]([C@@H](O)C([O-])=O)C([O-])=O
IUPAC Name(2R,3R)-2,3-dihydroxybutanedioic acid;sodium;dihydrate
Molecular Weight (g/mol)230.08
PubChem CID131855972
Formula Weight230.08
Percent Purity99.0 to 101.0%
Physical FormPowder or Solid
Unit SizeEach