Trifluoroacetic Acid (Peptide Synthesis Grade), Fisher BioReagents™
Trifluoroacetic Acid (Peptide Synthesis Grade), Fisher BioReagents™
Trifluoroacetic Acid (Peptide Synthesis Grade), Fisher BioReagents™
Fisher Bioreagents

Trifluoroacetic Acid (Peptide Synthesis Grade), Fisher BioReagents™

For peptide synthesis and sequencing protocols
Have Questions?
Catalog NumberQuantityPackaging
YBP618500500 mLAmber Glass
Catalog number YBP618500
HSN 29159090
Price (INR)
52,911.00
Each
Quantity:
500 mL
Packaging:
Amber Glass
Request bulk or custom format
Price (INR)
52,911.00
Each
Specifications
Absorbance0.06 max. at 275nm, 0.03 max. at 300nm
Chemical Name or MaterialTrifluoroacetic Acid
pH0.45
CAS76-05-1
Melting Point-15°C
View more
Trifluoroacetic acid is used in peptide sythesis and sequencing protocols.