New Zenon Alexa Fluor Plus Labeling Reagents are superior in three ways: improved bioconjugation for brighter signal, redesigned protein for robust screening regardless of IgG isotype, and new, more sensitive fluorophores.
Zenon&trade; Mouse IgG<sub>2a</sub> Labeling Kits
Invitrogen™

Zenon™ Mouse IgG2a Labeling Kits

Quickly generate antibody isotype conjugates for immunocytochemistry (ICC), immunohistochemistry (IHC), flow cytometry, and cell imaging with the Invitrogen Zenon™ mouse IgG2a Labeling Kits.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantityExcitation/EmissionLabel or Dye
Z2510850 Reactions kit650/668 nmAlexa Fluor 647
Z2510250 Reactions kit495/519 nmAlexa Fluor 488
Z2510750 Reactions kit590/617 nmAlexa Fluor 594
Z2515125 Reactions kit650/660 nmAPC (Allophycocyanin)
Z2515525 Reactions kit496, 546, 565/578 nmR-PE (R-Phycoerythrin)
Catalog number Z25108
Price (INR)
45,468.00
1 kit
Quantity:
50 Reactions kit
Excitation/Emission:
650/668 nm
Label or Dye:
Alexa Fluor 647
Price (INR)
45,468.00
1 kit
Quickly generate antibody isotype conjugates for immunocytochemistry (ICC), immunohistochemistry (IHC), flow cytometry, and cell imaging with the Invitrogen Zenon™ mouse IgG2a Labeling Kits. Simplify your laboratory applications and reduce antibody cross-reactivity while also achieving efficient labeling of antibodies in serum, ascites fluid, or hybridoma suspensions.

Achieve fast, versatile, and reliable fluorophore-, biotin-, or enzyme-labeled IgG2a primary antibodies with the Zenon™ mouse IgG2a labeling kits. These kits utilize Alexa Fluor fluorophores, biotin, Pacific Blue, or enzymes such as R-phycoerythrin and allophycocyanin, which are attached to monovalent, affinity purified Fab fragments. The Fab fragments, in turn, are directed against and bind with the Fc portion of IgG2a primary antibodies. Only a small amount of starting material is required, and the method is optimized for efficient labeling of antibodies in serum, ascites fluid, or hybridoma suspensions. Because the Zenon labeling method is based on immunoselectivity, it does not require the removal of exogenous proteins (such as serum) or amine-containing buffers from the target antibody, simplifying the process.

Zenon labeling technology greatly simplifies the use of multiple mouse-derived antibodies in the same staining protocol. Zenon tricolor labeling kits contain sufficient materials for 10 labeling reactions of each of three different fluorescent colors.

Important features of Zenon labeling technology:
• Labeled antibodies are typically ready to use in 10 minutes
• Requires only 1–20 μg primary antibody
• Simple—no purification required
• Flexible—choose from different fluorophores, biotin, HRP, alkaline phosphatase
• Multiplex with other mouse monoclonal antibodies simultaneously
• Can be used in a variety of applications including ICC, IHC, flow cytometry, and cell imaging.

Advantages of using Zenon antibody labeling kits:

Cost savings
Zenon antibody labeling kits offer a cost-conscious and reproducible method of tagging as little as 0.4 μg in 2 μL of primary antibody, with minimal waste of expensive or difficult-to-obtain antibodies, or excessive washing steps that pose the risk of product loss.

Sensitivity
Label your primary antibodies without compromising their antigen binding affinity: Zenon dye- and enzyme-labeled Fab fragments, which are targeted to the Fc tail, are affinity purified during their preparation to ensure high affinity and selectivity for the Fc portion of the corresponding primary antibody. The procedure for chemical labeling of the Zenon Fab fragments protects their Fc-binding site, resulting in more active labeling reagents.

Speed
No purification procedure is required prior to using Zenon Fab fragments in your laboratory applications. Formation of the Fab-antibody complex occurs in fewer than five minutes, followed by a five-minute blocking step. During this time, almost all the primary antibody in the mixture is labeled with the labeled Fab fragments.

Simplicity
The Fab-antibody complexes display fluorescence or enzymatic activity that is similar in intensity to that of directly labeled primary antibodies. Varying the extent of the antibody labeling is as simple as changing the amount of added Zenon labeling reagent during the reaction. Once the labeling complexes are formed, they can used immediately, without need for antibody purification.

Reliability
The Zenon Fab-antibody complex is stable and allows subsequent or simultaneous labeling of different target cells and tissues with different complexes. After staining, an aldehyde-based fixing step may be used to prevent the transfer of different labels between different primary antibodies, preserving the initial staining pattern.

Customization
We offer custom antibody conjugation services that are efficient and confidential, and we stand by the quality of our work. We are ISO 13485:2000 certified.

For Research Use Only. Not for use in diagnostic procedures.
Specifications
ColorFar-Red
Excitation/Emission650/668 nm
Label TypeAlexa Fluor
Labeling Scale< 1–20 μg
Product LineZenon
Product TypeLabeling Kit
Quantity50 Reactions kit
SpeciesMouse
Labeling TargetIgG2a
Label or DyeAlexa Fluor 647
Unit Size1 kit
Contents & Storage
Contains 1 vial of Zenon Alexa Fluor 647 mouse IgG2a labeling reagent (250 μL), and 1 vial of Zenon blocking reagent (mouse IgG, 250 μL).

Store in refrigerator (2–8°C) and protect from light.

Citations & References (2)

Citations & References
Abstract
The maturation of mucus-secreting gastric epithelial progenitors into digestive-enzyme secreting zymogenic cells requires Mist1.
Authors:Ramsey VG, Doherty JM, Chen CC, Stappenbeck TS, Konieczny SF, Mills JC,
Journal:Development
PubMed ID:17164426
Continuous regeneration of digestive enzyme (zymogen)-secreting chief cells is a normal aspect of stomach function that is disrupted in precancerous lesions (e.g. metaplasias, chronic atrophy). The cellular and genetic pathways that underlie zymogenic cell (ZC) differentiation are poorly understood. Here, we describe a gene expression analysis of laser capture microdissection ... More
One-step analysis of protein complexes in microliters of cell lysate using indirect immunolabeling & fluorescence cross-correlation spectroscopy.
Authors:Stoevesandt O, Brock R
Journal:Nat Protoc
PubMed ID:17406236
This protocol describes a 'mix-and-measure' procedure for the analysis of interactions of endogenous proteins in microliters of crude cell lysates. The proteins of interest are labeled by indirect immunofluorescence through simple addition of all primary and secondary antibodies to the lysate. Detection is based on fluorescence cross-correlation spectroscopy. Due to ... More