Zymosan A <i>S. cerevisiae </i>BioParticles&trade;, Texas Red&trade; conjugate
Invitrogen™

Zymosan A S. cerevisiae BioParticles™, Texas Red™ conjugate

The Molecular Probes™ BioParticles™ product line consists of a series of fluorescently labeled, heat- or chemically killed bacteria and yeastRead more
Have Questions?
Catalog NumberQuantity
Z284310 mg
Catalog number Z2843
Price (INR)
-
Quantity:
10 mg
The Molecular Probes™ BioParticles™ product line consists of a series of fluorescently labeled, heat- or chemically killed bacteria and yeast in a variety of sizes, shapes, and natural antigenicities. These fluorescent BioParticles™ products have been employed to study phagocytosis by fluorescence microscopy, quantitative spectrofluorometry, and flow cytometry.

We offer E. coli (K-12 strain), S. aureus (Wood strain, without protein A) and zymosan (S. cerevisiae) BioParticles™ conjugates covalently labeled with a variety of different fluorophores (special care has been taken to remove free dye after conjugation). Unlike the fluorescence of fluorescein-labeled BioParticles™ conjugates, which is partially quenched in acidic environments, the fluorescence of the Alexa Fluor™, BODIPY™ FL, tetramethylrhodamine and Texas Red™ dye conjugates is uniformly intense over the pH range from 3 to 10.

BioParticles Specifications:
• Label (Ex/Em): Texas Red™ (∼595/615 nm)
• Particle: Zymosan (S. cerevisiae)
• Opsonizing reagent available


Using BioParticles Products
BioParticles™ conjugates are provided as lyophilized powders. There are approximately 3 x 108 E. coli or S. aureus particles per mg solid and approximately 2 x 107 zymosan particles per mg solid. BioParticles™ conjugates can be reconstituted in the buffer of your choice for use in phagocytosis assays. The fluorescence of BioParticles™ conjugates that are bound to the surface of the cell (but not internalized) can be quenched by ethidium bromide, trypan blue, or other quenchers. In addition to cellular applications, fluorescent BioParticles™ conjugates may be effective as flow cytometry calibration references when sorting bacteria and yeast mutants. These small particles may also be useful references for light scattering studies because their sizes and shapes differ in characteristic ways.

Find More BioParticles™ Products
We offer a large range of dye-labeled and unlabeled E. coli (K-12 strain), S. aureus (Wood strain, without protein A), and zymosan (S. cerevisiae) BioParticles™ products. Find out about these products and their applications by reviewing Probes for Following Receptor Binding and Phagocytosis—Section 16.1 in the Molecular Probes™ Handbook.

For pH-sensitive endocytosis assays, see our pHrodo™ BioParticles™ conjugates.

For Research Use Only. Not for human or animal therapeutic or diagnostic use.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Detection MethodFluorescence
Dye TypeTexas Red™
FormLyophilized Powder
Quantity10 mg
Shipping ConditionRoom Temperature
SpeciesS. cerevisiae
For Use With (Equipment)Fluorescence Microscope
Product LineBioParticles, Texas Red
Product TypeBioparticle Conjugate
pH3 to 10
Unit Size10 mg
Contents & Storage
Store in freezer (-5 to -30°C) and protect from light.

Frequently asked questions (FAQs)

Are the Invitrogen BioParticles products sterile?

While the bacteria have been attenuated with formaldehyde and alcohol desiccation, the BioParticles products are not considered sterile, and we do not recommend incubation of more than 4 hours. This applies to all of our dye-labeled (pHrodo, Alexa Fluor, etc.) and unlabeled BioParticles products.

What is the type of bond that attaches the dyes to the BioParticles probes?

We use amine-reactive dyes to covalently attach fluorescent dyes to all of our BioParticles probes such as the Escherichia coli (K-12 strain) BioParticles probes, Staphylococcus aureus (Wood strain without protein A) BioParticles, and the Zymosan A (S. cerevisiae) BioParticles probes.

Find additional tips, troubleshooting help, and resources within our Cell Analysis Support Center.

Citations & References (7)

Citations & References
Abstract
Kruppel-like factor 2 (KLF2) regulates proinflammatory activation of monocytes.
Authors:Das H,Kumar A,Lin Z,Patino WD,Hwang PM,Feinberg MW,Majumder PK,Jain MK
Journal:Proceedings of the National Academy of Sciences of the United States of America
PubMed ID:16617118
The mechanisms regulating activation of monocytes remain incompletely understood. Herein we provide evidence that Kruppel-like factor 2 (KLF2) inhibits proinflammatory activation of monocytes. In vitro, KLF2 expression in monocytes is reduced by cytokine activation or differentiation. Consistent with this observation, KLF2 expression in circulating monocytes is reduced in patients with ... More
Importance of MEK in neutrophil microbicidal responsiveness.
Authors:Downey GP, Butler JR, Tapper H, Fialkow L, Saltiel AR, Rubin BB, Grinstein S
Journal:J Immunol
PubMed ID:9552001
'Exposure of neutrophils to inflammatory stimuli such as the chemoattractant FMLP leads to activation of responses including cell motility, the oxidative burst, and secretion of proteolytic enzymes. A signaling cascade involving sequential activation of Raf-1, mitogen-activated protein kinase (MEK), and extracellular signal regulated kinase (ERK) is also rapidly activated after ... More
Myosin light chain phosphorylation does not increase during yeast phagocytosis by macrophages.
Authors:de Lanerolle P, Gorgas G, Li X, Schluns K
Journal:J Biol Chem
PubMed ID:8349578
'We have studied the role of myosin II light chain phosphorylation in yeast phagocytosis by J774 cells. J774 cells, which are mouse cells of monocyte/macrophage lineage, ingest opsonized yeast particles, and the rate of internalization is linear for 60 min at 37 degrees C. Immunoprecipitation of myosin II from cells ... More
Ca2+ and synaptotagmin VII-dependent delivery of lysosomal membrane to nascent phagosomes.
Authors:Czibener C, Sherer NM, Becker SM, Pypaert M, Hui E, Chapman ER, Mothes W, Andrews NW
Journal:J Cell Biol
PubMed ID:16982801
Synaptotagmin (Syt) VII is a ubiquitously expressed member of the Syt family of Ca2+ sensors. It is present on lysosomes in several cell types, where it regulates Ca2+-dependent exocytosis. Because [Ca2+]i and exocytosis have been associated with phagocytosis, we investigated the phagocytic ability of macrophages from Syt VII-/- mice. Syt ... More
Polyethylene glycol-modified GM-CSF expands CD11b(high)CD11c(high) but notCD11b(low)CD11c(high) murine dendritic cells in vivo: a comparative analysis with Flt3 ligand.
Authors:Daro E, Pulendran B, Brasel K, Teepe M, Pettit D, Lynch DH, Vremec D, Robb L, Shortman K, McKenna HJ, Maliszewski CR, Maraskovsky E
Journal:J Immunol
PubMed ID:10861034
Dendritic cells (DC) are potent APCs that can be characterized in the murine spleen as CD11b(high)CD11c(high) or CD11b(low)CD11c(high). Daily injection of mice of Flt3 ligand (FL) into mice transiently expands both subsets of DC in vivo, but the effect of administration of GM-CSF on the expansion of DC in vivo ... More