Product Image
Invitrogen™

ZOOM™ Carrier Ampholytes pH 9-11

ZOOM® Carrier Ampholytes are small, soluble molecules with both positive and negative charge groups. They sort based on their isoelectricRead more
Have Questions?
Catalog NumberQuantity
ZM002410 mL
Catalog number ZM0024
Price (INR)
34,036.00
Online offer
Ends: 31-Dec-2026
48,622.00
Save 14,586.00 (30%)
10 mL
Quantity:
10 mL
Price (INR)
34,036.00
Online offer
Ends: 31-Dec-2026
48,622.00
Save 14,586.00 (30%)
10 mL
ZOOM® Carrier Ampholytes are small, soluble molecules with both positive and negative charge groups. They sort based on their isoelectric points in an electric field, thereby establishing a pH gradient in solution or contributing to the gradient when used with IPG strips. Carrier ampholytes help stabilize the pH gradient and current in IPG strips and aid in protein solubility, resulting in reproducible IEF resolution.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Quantity10 mL
Shelf Life1 Year
Shipping ConditionRoom Temperature
Product LineZOOM
Product TypeCarrier Ampholyte
pH RangepH 9-11
Unit Size10 mL
Contents & Storage
ZOOM™ Carrier Ampholytes are supplied as 10 ml bottles. Store at +4°C. Guaranteed stable for 1 year when properly stored.

Frequently asked questions (FAQs)

What may cause streaking on the 2nd dimension gel after IEF?

There are several reasons why streaking may occur.

(1) Sample is not completely solubilized prior to application.

(2) Sample is poorly soluble in rehydration solution.

(3) Non-protein impurities in the sample can interfere with IEF, causing horizontal streaking in the final 2-D result, particularly toward the acidic side of the gel.

(4) Ionic impurities are present in sample.

(5) Ionic detergent is present in sample.

(6) Sample load is too high.

(7) Underfocusing. Focusing time was not long enough to achieve steady state focusing.

(8) Overfocusing. Extended focusing times (over 100,000 Vh) may result in electroendosmotic water and protein movement, which can produce horizontal smearing.

What should be done?

(1) Be sure that the sample is completely and stably solubilized. Note: Repeated precipitation-resolubilization cycles produce or increase horizontal streaking.

(2) Increase the concentration of the solubilizing components in the rehydration solution.

(3) Modify sample preparation to limit these contaminants or dialyze protein.

(4) Reduce salt concentration to below 10 mM by dilution or desalt the sample by dialysis. Precipitation with TCA and acetone and subsequent resuspension is another effective desalting technique that removes lipids, nucleotides and other small molecules.
Note: Specific and non-specific losses of proteins can occur with dialysis, gel chromatography, and precipitation/resuspension of samples. If the sample preparation cannot be modified, the effect of ionic impurities can be reduced by modifying the IEF protocol. Limit the voltage to 100-150 V for 2 hours, then resume a normal voltage step program. This pre-step allows the ions in the sample to move to the ends of the IPG strip.

(5) If the ionic detergent SDS is used in sample preparation, the final concentration must not exceed 0.25% after dilution into the rehydration solution. Additionally, the concentration of the non-ionic detergent present must be at least 8 times higher than the concentration of any ionic detergent to ensure complete removal of SDS from the proteins.

(6) Extend focusing time. Load less sample.

(7) Prolong focusing time.

(8) Reduce focusing time.

Find additional tips, troubleshooting help, and resources within our Protein Assays and Analysis Support Center.

When running the ZOOM IEF Fractionator, if there is not a solubility issue, will the proteins/peptides in the sample migrate to their expected chambers based on isoelectric point in the absence of carrier ampholytes? Won't they act as ampholytes within each chamber?

Proteins are ampholytes, but in general they are poor carrier ampholytes. The profiles of the fractions are similar, but not identical, to those obtained with ampholytes in the sample.

Find additional tips, troubleshooting help, and resources within our Protein Assays and Analysis Support Center.

Can I heat my protein sample prepared using ZOOM 2D Protein Solubilizer?

We do not recommend heating protein samples containing urea over 37 degrees C as elevated temperatures cause urea to hydrolyze to isocyanate, which modifies proteins by carbamylation.

Find additional tips, troubleshooting help, and resources within our Protein Electrophoresis and Western Blotting Support Center.

How do you recommend storing protein samples prepared in the Sample Rehydration buffer?

We recommend storing them at -80 degrees C. We do not recommend storing them at -20 degrees C

Find additional tips, troubleshooting help, and resources within our Protein Electrophoresis and Western Blotting Support Center.

How much ampholyte do you recommend adding to the sample rehydration buffer?

The recommended ampholyte concentration in the sample rehydration buffer is 0.5%.

*If you are loading 5-50 µg of protein (pure protein or crude lysate) per ZOOM strip, use 0.5% ampholytes in the sample rehydration buffer.
*If you are loading >50 µg of protein (crude lysate or fractionated sample) per ZOOM Strip, use 0.5-2% ampholytes in the sample rehydration buffer.

Note: Higher ampholyte concentration requires longer focusing times.

Find additional tips, troubleshooting help, and resources within our Protein Electrophoresis and Western Blotting Support Center.

Citations & References (1)

Citations & References
Abstract
Fluorescent carrier ampholytes assay for portable, label-free detection of chemical toxins in tap water.
Authors:Bercovici M, Kaigala GV, Backhouse CJ, Santiago JG,
Journal:Anal Chem
PubMed ID:20141152
We present a novel method for fluorescence-based indirect detection of analytes and demonstrate its use for label-free detection of chemical toxins in a hand-held device. We fluorescently label a mixture of low-concentration carrier ampholytes and introduce it into an isotachophoresis (ITP) separation. The carrier ampholytes provide a large number of ... More