Product Image
Applied Biosystems™
Pathatrix™ Auto Plastics Kit
Have Questions?
Catalog NumberQuantity
ZNACPT50 Pack
Catalog number ZNACPT
Price (INR)
-
Quantity:
50 Pack
A box of 50 sterile, single use plastic consumable kits for use with the Pathatrix™ Auto Instrument, no paramagnetic particles included. The total working volume is 10 – 60 ml. Each pack includes sufficient sterile single use plastic consumable kits for 50 tests.
Specifications
Capacity10 to 60 mL sample volumes
For Use With (Equipment)Pathatrix™ Auto System
Product LinePathatrix
Quantity50 Pack
Shipping ConditionRoom Temperature
Unit Size50 pack
Contents & Storage
50 sterile, single use plastic assemblies. Includes sample vessels, elution vessels, and capture phase assemblies.
Store kit at room temperature.