Nicolet™ iN10 Infrared Microscope
Nicolet™ iN10 Infrared Microscope
Nicolet™ iN10 Infrared Microscope
Nicolet™ iN10 Infrared Microscope
Nicolet™ iN10 Infrared Microscope
Nicolet™ iN10 Infrared Microscope
Nicolet™ iN10 Infrared Microscope
Thermo Scientific™

Nicolet™ iN10 Infrared Microscope

Streamline your infrared microanalysis workflow with the Thermo Scientific™ Nicolet™ iN10 Infrared Microscope.

Have Questions?
Catalog NumberFor Use With (Application)
IQLAADGAAGFAHDMAPDMicrospectroscopy, Material Identification, Packaging and Laminates, Coatings, API Mixtures Distribution Mapping, Contaminant Identification, Failure Analysis
Catalog number IQLAADGAAGFAHDMAPD
Price (USD)
-
Request A Quote
For Use With (Application):
Microspectroscopy, Material Identification, Packaging and Laminates, Coatings, API Mixtures Distribution Mapping, Contaminant Identification, Failure Analysis

Streamline your infrared microanalysis workflow with the Thermo Scientific™ Nicolet™ iN10 Infrared Microscope. Designed with the simplicity of an FTIR spectrometer and the ability to measure samples down to a few microns, it is a modern, productive and cost-effective infrared microanalysis solution. The Nicolet iN10 Infrared Microscope provides answers with the high degree of confidence required by today's analytical laboratories.

With its intuitive and innovative optical design, combined with its novel approach to instrument operation, the Nicolet iN10 Infrared Microscope allows users of all skill levels to easily and confidently perform their microanalysis functions.

The Nicolet iN10 Infrared Microscope Features:

  • Thermo Scientific™ OMNIC™ Specta Software provides spectral identification of pure compounds and mixtures
  • Integrated design eliminates need for external spectrometer
  • Compact footprint saves laboratory space and budget
  • Room temperature detector eliminates need for liquid nitrogen for samples down to 50-microns
  • Computer-controlled automation allows access to all system settings from the computer
  • Programmable sample coordinates reduce time required for repetitive tasks
  • Dual monitor operation offers maximum viewing
  • Fully automated system performance verification provides maximum confidence

Configurable Options

  • Direct contact sampling with MicroTip ATR
  • Sensitivity enhancement with LN-cooled MCT detector
  • Manual or motorized stage
  • Enhanced visual inspection with motorized visible polarizer
  • Expand with Thermo Scientific™ Nicolet™ iZ10 FT-IR Module to work with standard sample-compartment accessories and full spectrometer capabilities

Ordering Alerts: This infrared microscope is sold as a configured system designed to meet your specific application needs. For more information, contact your Thermo Scientific Sales Representative.

Recommended for:

  • Microspectroscopy
  • Material identification
  • Packaging and laminate
  • Coating
  • API mixture distribution mapping
  • Contaminant identification
  • Failure analysis
Specifications
Amperage3.2A
Certifications/ComplianceCE, ETL, Manufactured in an ISO 9001:2000 certified facility
Depth (Metric)653 mm
DescriptionNicolet iN10 Infrared Microscope
Detector TypeStandard: DTGS; Optional: MCT-A
Frequency47/63 Hz
Height (Metric)533 mm
IncludesSampling Tool Kit
Voltage100/240 VAC
Warranty1 year full warranty, complete system
Width (Metric)622 mm
For Use With (Application)Microspectroscopy, Material Identification, Packaging and Laminates, Coatings, API Mixtures Distribution Mapping, Contaminant Identification, Failure Analysis
ModelNicolet iN10 Infrared Microscope
Unit SizeEach