Copper(II) oxide, 99.7% (metals basis)
Copper(II) oxide, 99.7% (metals basis)
Copper(II) oxide, 99.7% (metals basis)
Copper(II) oxide, 99.7% (metals basis)
Thermo Scientific Chemicals

Copper(II) oxide, 99.7% (metals basis)

CAS: 1317-38-0 | CuO | 79.55 g/mol
Quantity:
100 g
2 kg
Catalog number 041692.22
also known as 041692-22
Price (JPY)
-
Request A Quote
Quantity:
100 g
Request bulk or custom format
Chemical Identifiers
CAS1317-38-0
IUPAC Namecopper(2+) oxidandiide
Molecular FormulaCuO
InChI KeyKKCXRELNMOYFLS-UHFFFAOYSA-N
SMILES[O--].[Cu++]
View more
SpecificationsSpecification SheetSpecification Sheet
Total Metal Impurities0.3% max.
Copper(II) oxide is used as an adsorbent and absorbent, pigment, antifouling agent, wood preservative, optical glass polishing agent, welding flux for bronze materials, agricultural chemical and abrasive. It is used in the preparation of intermediates, oxidizing/reducing agents, surface treating agents and process regulators. It finds use in the manufacture of rayons, phosphors, and dry battery cells. It serves as a catalyst in certain chemical reactions of industrial importance. It is also employed in chemical sensors and superconducting materials. Hazardous materials like cyanides, dioxins and halogenated hydrocarbons can be disposed through oxidation with copper oxide.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only