Search
| Catalog Number | Maximum Pressure | Air Displacement |
|---|---|---|
| 01257156 | 40 torr | 35 L/min. |
Designed for use with rotary evaporators, concentrators and vacuum ovens.
Use these oil-free DryFast vacuum pumps for applications requiring chemical resistance. DryFast pumps provide mid-range vacuum (2 to 79 torr) with easy adjustment and low maintenance. Flow rates from 25 to 70 L/min. Select a DryFast model to suit your application needs:
Applications
Design Advantages