Specifications
Certifications/ComplianceNFPA 99
Collar TypeNotched Lapel
Construction1.6 oz. Fabric
For Use With (Application)Laboratories, Remediation, Decontamination, Pesticides
Length (English)38.5 in.
Length (Metric)97.79 cm
Model10029
Seam TypeSerged
Size (English) Chest39 to 41 in.
Closure TypeSnaps
ColorWhite
Cuff StyleOpen
DisposableLimited-use
GenderUnisex
MaterialSMS
No. of Pockets2
Product TypeGeneral Purpose Lab Coat
SizeLarge
SterilityNon-sterile
StyleKnee-length
Unit SizeCase of 25