Tyvek™ 400 Bib Apron
Fisher Scientific™
Tyvek™ 400 Bib Apron
Nonwoven Tyvek fabric helps resist penetration by particles and other contaminants, inherent barrier stops submicron particles even after abrasion
Have Questions?
Catalog number 0136183TP
also known as FISHERMIS-0136183-100/CS
Price (KRW)
-
Nonwoven Tyvek fabric helps resist penetration by particles and other contaminants. The inherent barrier stops submicron particles even after abrasion.
  • Provides enhanced frontal protection
  • Bound seams around neck
  • Ties at waist
  • Manufactured under specifications that do not contain natural rubber latex
  • One size fits most
  • D13398751, D15080185
Specifications
For Use With (Application)Lead and Asbestos Abatement/Remedation, General Maintenance/Operations, Spray Painting, General Clean-up
Length (English)36 in.
Length (Metric)69 cm
Seam TypeBound
Closure TypeWaist Ties
ColorWhite
MaterialTyvek
SizeUniversal
TypeBib Apron
Unit SizeCase of 100