Biohazard Specimen Transport Bags
Biohazard Specimen Transport Bags
Fisher Scientific™

Biohazard Specimen Transport Bags

For light-duty use
Have Questions?
Change viewbuttonViewtableView
Catalog NumberDimensions (L x W)Includes
0180001
also known as FIS-0180001
6 x 6 in. (15.24 x 15.24cm)Document compartment
0180000
also known as FIS-0180000
4 x 6 in. (10.16 x 15.24cm)Document compartment
0180004
also known as FIS-0180004
9 x 6 in. (22.86 x 15.24cm)Requisition Pouch, STAT label
0180005
also known as FIS-0180005
9 x 6 in. (22.86 x 15.24cm)3 x 5 in. Absorbent Pad
0180006
also known as FIS-0180006
6 x 6 in. (15.24 x 15.24cm)Pouch with flap, Scoreline
0180007
also known as FIS-0180007
8 x 8 in. (20.3 x 20.32cm)Document compartment
0180008
also known as FIS-0180008
10 x 8 in. (25.4 x 20.32cm)Document compartment
0180009
also known as FIS-0180009
12 x 15 in. (30.48 x 38cm)Document compartment
0180003
also known as FIS-0180003
9 x 6 in. (22.86 x 15.24cm)Document compartment
Catalog number 0180001
also known as FIS-0180001
Price (KRW)
-
Dimensions (L x W):
6 x 6 in. (15.24 x 15.24cm)
Includes:
Document compartment
For light-duty use
  • Clear polyethylene with three-wall construction and large printed biohazard graphic
  • Reclosable zipper seal
  • Cat. Nos. 01-800-02, 01-800-05 and 01-800-06 feature attached document compartment with flap cover that keeps paperwork in place
  • Cat. Nos. 01-800-00, 01-800-01, 01-800-03, 01-800-04, 01-800-07, 01-800-08, and 01-800-09 feature attached document compartment without flap cover
  • Cat. No. 01-800-05 features a pouch and a 3 x 5 in. (7.6 x 5.1cm) pad impregnated with superabsorbent polymers; pad will absorb 180cc of water, 60cc of urine or 38cc of blood
  • Cat. No. 01-800-06 features a scoreline for safe and easy entry without cutting
Specifications
Capacity (English)1 lb.
Certifications/ComplianceOSHA, NCCLS
Dimensions (L x W)6 x 6 in. (15.24 x 15.24cm)
For Use With (Application)For light-duty use
IncludesDocument compartment
Length (Metric)15.24 cm
No. per Case1000
No. per Pack100
Width (Metric)15.24 cm
AutoclavableNot Autoclavable
Closure TypeZip Closure
ColorClear
Length (English)6 in.
MaterialPolyethylene
Product TypeBiohazard Specimen Transport Bag
Width (English)6 in.
Unit SizeCase of 1000