Autoclavable HD Biohazard Waste Bags
Fisher Scientific™

Autoclavable HD Biohazard Waste Bags

Biohazard symbol printed bags to ensure safe handling and disposal of biohazardous waste.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberDimensions (L x W)
01815B
also known as FISHERMIS-01815B-100/CS
30 x 24 in.
01815A
also known as FIS-01815A
24 x 12 in.
01815C35 x 24 in.
Catalog number 01815B
also known as FISHERMIS-01815B-100/CS
Price (KRW)
-
Dimensions (L x W):
30 x 24 in.
Impact- and tear-resistant autoclave bags for waste sterilization. Fisherbrand™ Orange Autoclave Bags without Sterilization Indicator are constructed of high-density polyethylene.
  • Impact- and tear-resistant.
  • Bright orange color.
  • High-density polyethylene construction
Specifications
Certifications/ComplianceASTM
Closure TypeNone
Dimensions (L x W)30 x 24 in.
IncludesPrinted in English
Length (English)30 in.
Length (Metric)76 cm
Temperature (Metric)127°C
Thickness38 micron
Width (English)24 in.
Width (Metric)61 cm
AutoclavableAutoclavable
ColorOrange with Black Print
MaterialHDPE
Unit SizeCase of 100