Autoclavable HD Biohazard Waste Bags
Fisher Scientific™

Autoclavable HD Biohazard Waste Bags

Autoclavable, polyethylene biohazard bags ensure safe handling and disposal of biohazardous waste.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberColorDimensions (L x W)Thickness
01827CClear with Red Print30 x 24 in. (76 x 61 cm)1.50 mil
018141Natural24 x 12 in. (61 x 30 cm)1.50 mil
018143
also known as FIS-018143
Natural24 x 36 in. (61 x 91 cm)1.50 mil
01827A
also known as FIS-01827A
Clear with Red Print12 x 8 in. (30 x 20 cm)1.50 mil
01827D
also known as FIS-01827D
Clear with Red Print36 x 24 in. (91 x 61 cm)1.50 mil
01827BClear with Red Print24 x 12 in. (61 x 30 cm)1.50 mil
01829A
also known as FISHERMIS-01829A-200/CS
Red with Black Print12 x 8 in. (30 x 20 cm)1.50 mil
01829B
also known as FISHERMIS-01829B-200/CS
Red with Black Print19 x 14 in. (48 x 36 cm)1.50 mil
01829CRed with Black Print23 x 19 in. (58 x 48 cm)1.50 mil
01829D
also known as FISHERMIS-01829D-200/CS
Red with Black Print25 x 35 in. (64 x 89 cm)1.50 mil
01829FRed with Black Print30 x 33 in. (76 x 84 cm)1.50 mil
01831A
also known as FISHERTOPSELLING-01831A-EACH
Red with Black Print36 x 24 in. (91 x 61 cm)4 mil
01831B
also known as FIS-01831B
Red with Black Print48 x 38 in. (122 x 97 cm)4 mil
01832AClear36 x 24 in. (91 x 61 cm)4 mil
01829ERed with Black Print48 x 38 in. (122 x 97 cm)1.50 mil
Catalog number 01827C
Price (KRW)
-
Color:
Clear with Red Print
Dimensions (L x W):
30 x 24 in. (76 x 61 cm)
Thickness:
1.50 mil
Autoclavable, polyethylene biohazard bags to ensure safe handling and disposal of biohazardous waste.
  • All bags supplied with autoclavable elastic steam-venting closures except 1.5 mil-thick plain clear bags
  • Polyethylene
  • Temperature range 260°F
  • Available in 4 colors/styles
Specifications
Certifications/ComplianceOSHA, 29 CFR1910.1030
Closure TypeElastic
Dimensions (L x W)30 x 24 in. (76 x 61 cm)
IncludesPrinted in English and Spanish
Length (English)30 in.
Length (Metric)76 cm
Temperature (Metric)127°C
Thickness1.50 mil
Width (English)24 in.
Width (Metric)61 cm
AutoclavableAutoclavable
ColorClear with Red Print
MaterialPolyethylene
Product TypeAutoclavable Bags
Unit SizeCase of 200