Search
| Catalog Number | Diameter (Metric) | For Use With (Equipment) |
|---|---|---|
| 02612A also known as FIS-02612A | 6.5 cm | For 150 mL beaker |
| 02612E also known as FIS-02612E | 12.5 cm | For 1000 mL beaker |
| 02612D also known as FISHERTOPSELLING-02612D-EACH | 10 cm | For 600 mL beaker |
These watch glasses possess high chemical durability, thus preventing contamination of solutions. They are molded to the same radius of curvature for convenient stacking. The heavy wall and uniform fire-polished edge provide mechanical strength. The outside diameter cannot be controlled within close limits and a slight degree of out-of-round can be expected to occur.