Polypropylene Microtiter Tubes
Fisher Scientific™

Polypropylene Microtiter Tubes

Ideal for sample dilution and/or mixing prior to transfer into microtiter plates.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberSterilityLabel TypePackaging
02681386SterileNo Writing AreaRobotic Pack
02681376
also known as FISHERTOPSELLING-02681376-EACH
Non-sterileNo Writing AreaBag
02681385Non-sterileNo Writing AreaRobotic Pack
Catalog number 02681386
Price (KRW)
-
Sterility:
Sterile
Label Type:
No Writing Area
Packaging:
Robotic Pack
Ideal for sample dilution and/or mixing prior to transfer into microtiter plates

Also suitable for HTLV-III testing, RIA, EIA, PCR, pharmaceutical quality control, sample freezer storage or transport of specimens and reagents. Choose plain tubes or tubes with frosted writing area. 

  • Superior-grade polypropylene tubes and packing racks are autoclavable and can be frozen to -80°C
  • Nonsterile tubes are certified RNase- and DNase-free
  • Sterile tubes are certified RNase-, DNase-, ATP-, DNA- and pyrogen-free
  • Frosted tubes feature writing area that wraps around tube collar
  • Capacity- 1.2mL
Specifications
Certifications/ComplianceRNase, DNase, ATP, DNA, pyrogen-free
For Use With (Application)HTLV-III testing, RIA, EIA, PCR, pharmaceutical quality control, sample freezer storage or transport of specimens and reagent
Label TypeNo Writing Area
PackagingRobotic Pack
Product TypeMicrotiter Tubes
AutoclavableAutoclavable
Capacity (Metric)1.2 mL
ColorNatural
GraduatedNo
MaterialPolypropylene
SterilitySterile
Unit SizePack of 960