You need to enable JavaScript to run this app.
Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Pipettes and Pipette Tips
Lab Centrifuges
Ultra-Low Temperature Freezers
Spectroscopy
Beakers
PCR Equipment and Supplies
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
See all product categories
Applications
Cell Analysis
Lab Equipment
Real-Time PCR
PCR
Chromatography
Cell Culture and Transfection
DNA and RNA Extraction and Analysis
Protein Biology
Flow Cytometry
Chemicals
See all applications and techniques
Services
Custom Services
Lab Informatics
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Instrument Services
Training Services
Unity Lab Services
See all services
Help and Support
Customer Center
Contact Us
Certificates of Analysis and Conformance
Safety Data Sheets (SDS)
Manuals
How to Cite Our Products in a Paper
Instrument Support
Knowledge Base and Product FAQs
Learning Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
Services Central
Home
▸
Shop All Products
▸
Pipette Products
▸
Pipette Tips
▸
Pipette Specific Tips
▸
Maxi Pipet Tips
Fisher Scientific™
Maxi Pipet Tips
For use with large volume pipets
Have Questions?
Change view
Catalog Number
Volume (Metric)
02681409
also known as PSELLING-02681409-PKFISHERTO
500 to 2500 μL
211953
also known as FISHERTOPSELLING-211953-EACH
1000 to 5000 μL
02707467
1 to 5 mL
02681410
5000 μL
02707131
250 to 2500 μL
5 Options
Catalog number
02681409
also known as PSELLING-02681409-PKFISHERTO
Price (KRW)
-
Volume (Metric):
500 to 2500 μL
For use with large volume pipets
Designed for use when transferring large sample volume
Advanced molding technology ensure tips are straight
Alternative option to serological pipets
Nonsterile
Certified by independent laboratory to be RNase, DNase, DNA, and Pyrogen free
Specifications
Graduated
No
Length (English)
4.5 in.
Length (Metric)
115 mm
Autoclavable
Autoclavable
Color
Natural
Filtered
Non-filtered
For Use With (Equipment)
Eppendorf Reference, Socorex
Format
Bulk
Material
Polypropylene
Sterility
Non-sterile
Tip Style
Macro
Volume (Metric)
500 to 2500 μL
Unit Size
Pack of 500