Aerosol-Barrier Tips
Fisher Scientific™

Aerosol-Barrier Tips

Prevent cross-contamination by using superior aerosol-barrier tips.

Have Questions?
Change viewbuttonViewtableView
Catalog NumberVolume (Metric)Format
0270766200 μL (Extended)Bulk
0270780200 μLRack
0270742200 μLHinged Rack
02707511000 to 1300 μLRack
0270768200 μL (Extended)Rack
0270777200 μL (Extended)Rack
027071671250 μLRack
027071681 to 5 mLBulk
Catalog number 0270766
Price (KRW)
-
Volume (Metric):
200 μL (Extended)
Format:
Bulk

Fisherbrand™ Aerosol-Barrier tips can be used universally and protects the intergrity of your samples.

  • Unique small micron filter prevents aerosol contamination with no obstruction to airflow
  • Use aerosol barrier tips to ensure result integrity
  • RNase, DNase, DNA, and pyrogen free
  • Natural color

Packaging Code -- BP: Bulk Pack; RI: Rack Inserts; HR: Hinged Rack; HRS: Hinged Rack, Sterile; HR/T: Hinged Rack, Trace Metal-free; LR: Lift-off Rack; LRS: Lift-off Rack, Sterile; SR: Stack Rack; IWS: Individually Wrapped, Sterile.

Specifications
Length (English)2.3 in.
Length (Metric)5.84cm
AutoclavableAutoclavable
ColorNatural
FilteredFiltered
For Use With (Equipment)Universal Fit
FormatBulk
SterilitySterile
Tip StyleExtended Length
Volume (Metric)200 μL (Extended)
Unit SizePack of 1000