You need to enable JavaScript to run this app.
Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Pipettes and Pipette Tips
Lab Centrifuges
Ultra-Low Temperature Freezers
Spectroscopy
Beakers
PCR Equipment and Supplies
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
See all product categories
Applications
Cell Analysis
Lab Equipment
Real-Time PCR
PCR
Chromatography
Cell Culture and Transfection
DNA and RNA Extraction and Analysis
Protein Biology
Flow Cytometry
Chemicals
See all applications and techniques
Services
Custom Services
Lab Informatics
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Instrument Services
Training Services
Unity Lab Services
See all services
Help and Support
Customer Center
Contact Us
Certificates of Analysis and Conformance
Safety Data Sheets (SDS)
Manuals
How to Cite Our Products in a Paper
Instrument Support
Knowledge Base and Product FAQs
Learning Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
Services Central
Home
▸
Shop All Products
▸
Pipette Products
▸
Pipette Tips
▸
Universal Pipette Tips
▸
Low-Retention Pipette Tips, Nonfilter
Fisher Scientific™
Low-Retention Pipette Tips, Nonfilter
Increase accuracy by reducing sample retention
Have Questions?
Change view
Catalog Number
Volume (Metric)
Sterility
02717149
1000 μL
Non-sterile
02717132
10 μL
Sterile
02717150
1000 μL
Non-sterile
02717151
1000 μL
Sterile
4 Options
Catalog number
02717149
Price (KRW)
-
Volume (Metric):
1000 μL
Sterility:
Non-sterile
Increase accuracy by reducing sample retention
Low retention surface minimizes samples loss
Certified by independent laboratory
Nonsterile: RNase, DNase, DNA, and Pyrogen Free; Sterile: RNase, DNase, DNA, Bioburden, PCR Inhibitor, and Pyrogen Free
Natural color
Specifications
Description
Low RetentionNon-sterile, Bulk
Model
Low Retention
Color
Natural
Filtered
Non-filtered
For Use With (Equipment)
Universal Fit
Format
Bulk
Sterility
Non-sterile
Tip Style
Extended Length
Volume (Metric)
1000 μL
Unit Size
Pack of 1000